102245
Casein Acid Hydrolysate for Biotechnology and Microbiology
Prime, for Biotechnology and Microbiology
Synonym(s):
2,6-Dimethyl-3-pyridinylamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H10N2
CAS Number:
Molecular Weight:
122.17
Beilstein:
110353
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Product Name
2-(Dimethylamino)pyridine, 97%
Assay
97%
refractive index
n20/D 1.559 (lit.)
bp
191 °C (lit.)
density
0.984 g/mL at 25 °C (lit.)
functional group
amine
SMILES string
CN(C)c1ccccn1
InChI
1S/C7H10N2/c1-9(2)7-5-3-4-6-8-7/h3-6H,1-2H3
InChI key
PSHKMPUSSFXUIA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
2-(Dimethylamino)pyridine is a clear, colorless to light yellow liquid.
Application
2-(Dimethylamino)pyridine in toluene at room temperature in the presence of 1.1 equivalent of methyl trifluoromethanesulfonate forms (2-pyridyl)-trimethylammonium trifluoromethanesulfonate salt. It is used as a model substrate in the preparation of chelate carbene via cyclometalation, H2 loss and reversible α-elimination.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Sagarika Samantray et al.
Nature communications, 11(1), 880-880 (2020-02-16)
A self-assembled coordination cage usually possesses one well-defined three-dimensional (3D) cavity whereas infinite number of 3D-cavities are crafted in a designer metal-organic framework. Construction of a discrete coordination cage possessing multiple number of 3D-cavities is a challenging task. Here we
2-, 3-and 4-[18F] Fluoropyridine by no-carrier-added nucleophilic aromatic substitution with K [18F] F-K222-a comparative study.
Karramkam M, et al.
Journal of Labelled Compounds & Radiopharmaceuticals, 46(10), 979-992 (2003)
Farideh Mahmoodzadeh et al.
International journal of biological macromolecules, 126, 517-524 (2018-12-30)
Conventional chemotherapy suffers lack of bioavailability, selectivity and multidrug resistance (MDR). Nano-sized drug delivery systems (DDS) is developing aimed to solve several limitations of conventional drug delivery systems. These systems have been offered for targeting tumor tissue due to their
Martien A Würdemann et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(36), 8090-8100 (2020-03-28)
Pyrazines are an underreported class of N-heterocycles available from nitrogen-rich biomass presenting an interesting functional alternative for current aromatics. In this work, access to pyrazines obtained from amino acids by using the 90 year old Dakin-West reaction was explored. After
Hong-Mei Liu et al.
International journal of nanomedicine, 12, 1065-1083 (2017-02-23)
Here, we report the hypoxia-responsive ionizable liposomes to deliver small interference RNA (siRNA) anticancer drugs, which can selectively enhance cellular uptake of the siRNA under hypoxic and low-pH conditions to cure glioma. For this purpose, malate dehydrogenase lipid molecules were
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service