Merck
  • Multi-class determination of undesirables in aquaculture samples by gas chromatography/tandem mass spectrometry with atmospheric pressure chemical ionization: A novel approach for polycyclic aromatic hydrocarbons.
Multi-class determination of undesirables in aquaculture samples by gas chromatography/tandem mass spectrometry with atmospheric pressure chemical ionization: A novel approach for polycyclic aromatic hydrocarbons.
Talanta (2017-06-13)
T Portolés, B Garlito, Jaime Nácher-Mestre, M H G Berntssen, J Pérez-Sánchez
ABSTRACT
In this work, a method for the analysis of 24 PAHs in 19 different matrices, including fish tissues, feeds and feed ingredients, has been developed using gas chromatography coupled to triple quadrupole tandem mass spectrometry with atmospheric pressure chemical ionization source (GC-APCI-MS/MS). The method is based on a modification of the unbuffered QuEChERS method, using freezing as an additional clean-up step and applying a 20-fold dilution factor to the final extract. The procedure was also tested for 15 pesticides and 7 polychlorinated biphenyl (PCB) congeners in order to widen the scope of the method. The excellent sensitivity and selectivity provided by GC-APCI-MS/MS allowed the dilution of the sample extracts and quantification using calibration with standards in solvent for all the 19 matrices tested. The developed method was evaluated at 2, 5 and 50ngg
Product Description
Supelco
Azoxystrobin, PESTANAL®, analytical standard
Supelco
Tebuconazole, PESTANAL®, analytical standard
Dibenzo[a,e]pyrene, BCR®, certified reference material
Sigma-Aldrich
Acenaphthylene-d8, ≥98 atom % D, 98% (CP)