Merck
  • Multiple routes of endocytic internalization of PDGFRβ contribute to PDGF-induced STAT3 signaling.
Multiple routes of endocytic internalization of PDGFRβ contribute to PDGF-induced STAT3 signaling.
Journal of cell science (2016-12-17)
Kamil Jastrzębski, Daria Zdżalik-Bielecka, Agnieszka Mamińska, Yannis Kalaidzidis, Carina Hellberg, Marta Miaczynska
ABSTRACT
Platelet-derived growth factor receptor β (PDGFRβ) is a receptor tyrosine kinase which upon activation by PDGF-BB stimulates cell proliferation, migration and angiogenesis. Ligand binding induces intracellular signaling cascades but also internalization of the receptor, eventually resulting in its lysosomal degradation. However, endocytic trafficking of receptors often modulates their downstream signaling. We previously reported that internalization of PDGFRβ occurs via dynamin-dependent and -independent pathways but their further molecular determinants remained unknown. Here we show that, in human fibroblasts expressing endogenous PDGFRβ and stimulated with 50 ng/ml PDGF-BB, ligand-receptor uptake proceeds via the parallel routes of clathrin-mediated endocytosis (CME) and clathrin-independent endocytosis (CIE). CME involves the canonical AP2 complex as a clathrin adaptor, while CIE requires RhoA-ROCK, Cdc42 and galectin-3, the latter indicating lectin-mediated internalization via clathrin-independent carriers (CLICs). Although different uptake routes appear to be partly interdependent, they cannot fully substitute for each other. Strikingly, inhibition of any internalization mechanism impaired activation of STAT3 but not of other downstream effectors of PDGFRβ. Our data indicate that multiple routes of internalization of PDGFRβ contribute to a transcriptional and mitogenic response of cells to PDGF.
Sigma-Aldrich
IgG from mouse serum, technical grade, ≥80% (SDS-PAGE), buffered aqueous solution
Sigma-Aldrich
Latrunculin A, from sea sponge, ≥85% (HPLC), waxy solid
Sigma-Aldrich
Rac1 Inhibitor, Rac1 Inhibitor, CAS 1177865-17-6, is a cell-permeable, reversible inhibitor of Rac1 GDP/GTP exchange. Interferes with the interaction between Rac1 and Rac-specific GEFs Trio and Tiam1 (IC₅₀ ~50 µM).
Sigma-Aldrich
ML 141, ≥98% (HPLC)
Sigma-Aldrich
Y-27632 dihydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Dynamin Inhibitor I, Dynasore, The Dynamin Inhibitor I, Dynasore, also referenced under CAS 304448-55-3, controls the biological activity of Dynamin. This small molecule/inhibitor is primarily used for Membrane applications.
Sigma-Aldrich
Rho Kinase Inhibitor, The Rho kinase inhibitor, CAS 872543-07-6, is a cell-permeable, highly specific, reversible, potent, and ATP-competitive inhibitor of Rho-associated kinase (ROCK; Ki = 1.6 nM).