Skip to Content
Merck
All Photos(1)

Documents

553502

Sigma-Aldrich

NSC23766 trihydrochloride

≥93% (HPLC), lyophilized, Rac1 inhibitor, Calbiochem®

Synonym(s):

Rac1 Inhibitor, NSC23766

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H38Cl3N7
CAS Number:
Molecular Weight:
530.96
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Rac1 Inhibitor, Rac1 Inhibitor, CAS 1177865-17-6, is a cell-permeable, reversible inhibitor of Rac1 GDP/GTP exchange. Interferes with the interaction between Rac1 and Rac-specific GEFs Trio and Tiam1 (IC₅₀ ~50 µM).

Quality Level

Assay

≥93% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

solubility

water: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C24H35N7.3ClH/c1-6-31(7-2)12-8-9-16(3)27-24-28-18(5)14-23(30-24)29-19-10-11-22-20(15-19)21(25)13-17(4)26-22;;;/h10-11,13-16H,6-9,12H2,1-5H3,(H2,25,26)(H2,27,28,29,30);3*1H

InChI key

CPUHORIUXPQCHW-UHFFFAOYSA-N

General description

A cell-permeable pyrimidine compound that specifically and reversibly inhibits Rac1 GDP/GTP exchange activity by interfering with the interaction between Rac1 and Rac-specific GEFs (guanine nucleotide exchange factors) Trio and Tiam1 (IC50 ~50 µM). Shown to effectively inhibit Rac1-mediated cellular functions in NIH3T3 and PC-3 cells (effective dose ~50 to 100 μM). Exhibits no effect on Cdc42 or RhoA activation and does not affect Rac1 interaction with BcrGAP or PAK1. Reduces TRAP-induced and collagen stimulated platelet aggregation (IC50 = 50 mM and 64 mM, respectively).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Rac1
Product does not compete with ATP.
Reversible: yes
Target IC50: ~50 µM against Rac1 GDP/GTP exchange activity; 50 mM and 64 mM against TRAP-induced and collagen stimulated platelet aggregation

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Dwivedi, S., et al. 2010. J. Translational Med.8, 128.
Desire, L., et al. 2005. J. Biol. Chem.280, 37516.
Gao, Y., et al. 2004. Proc. Natl. Acad. Sci. USA101, 7618.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Shruthi Sriramkumar et al.
PloS one, 17(8), e0271584-e0271584 (2022-08-04)
Ovarian cancer (OC) is a lethal gynecological malignancy with a five-year survival rate of only 46%. Development of resistance to platinum-based chemotherapy is a common cause of high mortality rates among OC patients. Tumor and transcriptomic heterogeneity are drivers of
Audrey Miller Williams et al.
eLife, 11 (2022-09-27)
For a group of cells to migrate together, each cell must couple the polarity of its migratory machinery with that of the other cells in the cohort. Although collective cell migrations are common in animal development, little is known about
Kimia Ghaffari et al.
PLoS genetics, 17(3), e1009402-e1009402 (2021-03-20)
Impaired formation of the intrahepatic biliary network leads to cholestatic liver diseases, which are frequently associated with autoimmune disorders. Using a chemical mutagenesis strategy in zebrafish combined with computational network analysis, we screened for novel genes involved in intrahepatic biliary
Kee-Beom Kim et al.
Cancer research, 82(22), 4219-4233 (2022-09-15)
WNT signaling represents an attractive target for cancer therapy due to its widespread oncogenic role. However, the molecular players involved in WNT signaling and the impact of their perturbation remain unknown for numerous recalcitrant cancers. Here, we characterize WNT pathway
SLIT2/ROBO1 signaling suppresses mTORC1 for organelle control and bacterial killing.
Bhosle, et al.
Life science alliance, 6 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service