Merck
  • Japanese encephalitis virus activates autophagy as a viral immune evasion strategy.
Japanese encephalitis virus activates autophagy as a viral immune evasion strategy.
PloS one (2013-01-16)
Rui Jin, Wandi Zhu, Shengbo Cao, Rui Chen, Hui Jin, Yang Liu, Shaobo Wang, Wei Wang, Gengfu Xiao
ABSTRACT
In addition to manipulating cellular homeostasis and survivability, autophagy also plays a crucial role in numerous viral infections. In this study, we discover that Japanese encephalitis virus (JEV) infection results in the accumulation of microtubule-associated protein 1 light chain 3-II (LC3-II) protein and GFP-LC3 puncta in vitro and an increase in autophagosomes/autolysosomes in vivo. The fusion between autophagosomes and lysosomes is essential for virus replication. Knockdown of autophagy-related genes reduced JEV replication in vitro, as indicated by viral RNA and protein levels. We also note that JEV infection in autophagy-impaired cells displayed active caspases cleavage and cell death. Moreover, we find that JEV induces higher type I interferon (IFN) activation in cells deficient in autophagy-related genes as the cells exhibited increased phosphorylation and dimerization of interferon regulatory factor 3 (IRF3) and mitochondrial antiviral signaling protein (MAVS) aggregation. Finally, we find that autophagy is indispensable for efficient JEV replication even in an IFN-defective background. Overall, our studies provide the first description of the mechanism of the autophagic innate immune signaling pathway during JEV infection.
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Millipore
SignalBoostImmunoreaction Enhancer Kit
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film