407207
SignalBoostImmunoreaction Enhancer Kit
Synonym(s):
immunodetection signal enhancer, signal enhancer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
usage
sufficient for 1600 cm2
sufficient for 20 miniblot(s)
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
detection method
chemiluminescent
colorimetric
fluorometric
shipped in
wet ice
storage temp.
2-8°C
General description
SignalBoost Immunoreaction Enhancer Kit is designed to enhance the desired signal in assays such as immunoblotting, dot blotting, and ELISA. This kit significantly improves sensitivity and discrimination when assessing protein targets of low immunoreactivity or low expression levels. Enhancement of signal is especially marked for low affinity antibodies. It replaces conventional antibody diluents with Solution 1, which is used to dilute primary antibodies and Solution 2, which is used to dilute secondary antibodies. No additional steps, materials, or increase of assay time is required.
• Increases signal: minimizes background resulting in high signal to noise ratios
• Versatile: can be used with nitrocellulose or PVDF membranes and is compatible with chemiluminescent or colorimetric detection systems
• Convenient: ready-to-use format, no dilution necessary
• Increases signal: minimizes background resulting in high signal to noise ratios
• Versatile: can be used with nitrocellulose or PVDF membranes and is compatible with chemiluminescent or colorimetric detection systems
• Convenient: ready-to-use format, no dilution necessary
SignalBoost enhances antigen-antibody reactions, yielding signals several times to several tens of times higher than those obtained with conventional methods. It has been verified that enhancement is particularly marked for relatively inactive antibodies. Compositions optimized for primary and secondary antibodies respectively minimize background and yield high S/N ratios. It can be used for various assays involving antigen-antibody reactions, such as immunoblotting, dot blotting and ELISA. Components do not affect the activity of enzymes used for labeling (peroxidase, alkaline phosphatase, etc.), allowing use in assays using any of these enzymes. It can also be used for detection of both fluorescence and coloration. For convenience, SignalBoost is supplied in a ready-to-use form, is at an appropriate concentration for use, and does not need to be diluted. Simply dilute antibodies with this product instead of conventional diluents to use. It does not lengthen the time required for the assay.
Components
Solution 1 for Primary Antibodies, Solution 2 for Secondary Antibodies, and a user protocol.
Warning
Toxicity: Standard Handling (A)
Principle
SignalBoost Immunoreaction Enhancer Solution is a reactive reagent for use in immunoassays, and was developed to resolve the problems of low sensitivity and high background often encountered during procedures such as immunoblotting and ELISA (enzyme-linked immunosorbent assay). It can be used in a range of immunoassays.
Physical form
Format: Liquid, Antibody Diluent
Storage and Stability
Upon receipt, store the entire contents of the kit at 4°C. Protect from light. Following initial use, this product is stable for up to 1 year at 4°C.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Skin Sens. 1
Storage Class Code
12 - Non Combustible Liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
British journal of pharmacology, 176(13), 2111-2130 (2019-03-09)
Amyotrophic lateral sclerosis (ALS) is a fatal neurodegenerative disease typically more common in males, implicating androgens in progression of both patients and mouse models. Androgen effects are mediated by androgen receptor which is highly expressed in spinal motor neurons and
Oncology reports, 43(4), 1267-1277 (2020-04-24)
Long non‑coding RNAs (lncRNAs) are involved in colorectal cancer (CRC) progression, however the mechanisms remain largely unknown. The present study aimed to reveal the role and possible molecular mechanisms of a new LNCRNA, LINC00858, in CRC. LINC00858 was increased in
F1000Research, 5, 2825-2825 (2017-04-14)
Sphingosine kinase 2 (SK2) is a ubiquitously expressed lipid kinase that has important, albeit complex and poorly understood, roles in regulating cell survival and cell death. In addition to being able to promote cell cycle arrest and apoptosis under certain
PloS one, 8(1), e52909-e52909 (2013-01-16)
In addition to manipulating cellular homeostasis and survivability, autophagy also plays a crucial role in numerous viral infections. In this study, we discover that Japanese encephalitis virus (JEV) infection results in the accumulation of microtubule-associated protein 1 light chain 3-II
Scientific reports, 11(1), 9255-9255 (2021-05-01)
Amyotrophic lateral sclerosis (ALS) is an adult-onset neurodegenerative disorder caused by loss of motor neurons. ALS incidence is skewed towards males with typically earlier age of onset and limb site of onset. The androgen receptor (AR) is the major mediator
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service