Merck
  • Methacrylated gelatin and mature adipocytes are promising components for adipose tissue engineering.
Methacrylated gelatin and mature adipocytes are promising components for adipose tissue engineering.
Journal of biomaterials applications (2015-05-29)
Birgit Huber, Kirsten Borchers, Günter Em Tovar, Petra J Kluger
ABSTRACT
In vitro engineering of autologous fatty tissue constructs is still a major challenge for the treatment of congenital deformities, tumor resections or high-graded burns. In this study, we evaluated the suitability of photo-crosslinkable methacrylated gelatin (GM) and mature adipocytes as components for the composition of three-dimensional fatty tissue constructs. Cytocompatibility evaluations of the GM and the photoinitiator Lithium phenyl-2,4,6-trimethylbenzoylphosphinate (LAP) showed no cytotoxicity in the relevant range of concentrations. Matrix stiffness of cell-laden hydrogels was adjusted to native fatty tissue by tuning the degree of crosslinking and was shown to be comparable to that of native fatty tissue. Mature adipocytes were then cultured for 14 days within the GM resulting in a fatty tissue construct loaded with viable cells expressing cell markers perilipin A and laminin. This work demonstrates that mature adipocytes are a highly valuable cell source for the composition of fatty tissue equivalents in vitro. Photo-crosslinkable methacrylated gelatin is an excellent tissue scaffold and a promising bioink for new printing techniques due to its biocompatibility and tunable properties.
Sigma-Aldrich
TissueFab® bioink , (Gel)ma -UV/365 nm
Sigma-Aldrich
TissueFab® bioink , Alg(Gel)ma -Vis/525 nm
Sigma-Aldrich
Methacrylic anhydride, contains 2,000 ppm topanol A as inhibitor, ≥94%
Sigma-Aldrich
TissueFab® bioink , (Gel)ma -VIS/405nm, low endotoxin
Sigma-Aldrich
TissueFab® bioink , Alg(Gel)ma -UV/365 nm
Sigma-Aldrich
TissueFab® bioink Alg(Gel)ma -UV/365 nm
Sigma-Aldrich
TissueFab® - low endotoxin GelMA-UV bioink, 0.2 μm filtered, suitable for 3D bioprinting applications
Sigma-Aldrich
TissueFab® bioink , (GelAlgHA)MA Vis/405 nm, low endotoxin
Sigma-Aldrich
TissueFab® bioink kit, (Gel)ma Laminin -UV/365 nm, low endotoxin
Sigma-Aldrich
TissueFab® bioink kit, Fibronectin-UV/365nm
Sigma-Aldrich
TissueFab® bioink kit, (Gel)ma Fibrin (UV/365), low endotoxin
Sigma-Aldrich
TissueFab® GelAlg − LAP Bioink, low endotoxin, 0.2 μm filtered, suitable for 3D bioprinting applications
Sigma-Aldrich
TissueFab® bioink kit, (Gel)ma Laminin -Vis/405 nm, low endotoxin
Sigma-Aldrich
TissueFab® bioink kit, (Gel)ma Fibrin (Vis/405), low endotoxin