Merck
  • Material Decomposition in Dual-Energy Computed Tomography Separates High-Z Elements From Iodine, Identifying Potential Contrast Media Tailored for Dual Contrast Medium Examinations.
Material Decomposition in Dual-Energy Computed Tomography Separates High-Z Elements From Iodine, Identifying Potential Contrast Media Tailored for Dual Contrast Medium Examinations.
Journal of computer assisted tomography (2015-08-22)
Tobias Fält, Marcus Söderberg, Johan Wassélius, Peter Leander
ABSTRACT
The aim of this study was to determine the potential of different high-Z elements to act as contrast media (CMs) alongside iodine (I) in dual-CM, dual-energy (DE) computed tomography examinations. Gadolinium (Gd), tantalum (Ta), wolfram (W), gold (Au), and bismuth (Bi) in addition to I were examined at all available kilovolt settings in a DE computed tomography scanner. Dual-energy ratios were calculated by dividing attenuation at low kilovolt by attenuation at high kilovolt. Dual-energy data sets were loaded into material decomposition software to evaluate separation of the elements from I. The DE ratios of Ta, W, and Au ranged between 0.9 and 1.2, being considerably lower than I at 1.9 to 2.6. These elements were completely separated from I using material decomposition. Gadolinium and Bi were more similar to I at 1.4 to 1.9. However, separation was nearly complete for Bi and suboptimal for Gd. Tantalum, W, and Au are ideal candidates for dual-CM examinations, whereas Bi is a slightly weaker candidate.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone, S15, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Tungsten(VI) chloride, ≥99.9% trace metals basis
Sigma-Aldrich
Tungsten(VI) chloride, powder, ≥99.99% trace metals basis (purity excludes molybdenum)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 2% 2-butanone, F25 MEK1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology