Merck
  • Survival of Salmonella during Drying of Fresh Ginger Root (Zingiber officinale) and Storage of Ground Ginger.
Survival of Salmonella during Drying of Fresh Ginger Root (Zingiber officinale) and Storage of Ground Ginger.
Journal of food protection (2015-11-12)
Dana R Gradl, Lingxiang Sun, Emily L Larkin, Stuart J Chirtel, Susanne E Keller
ABSTRACT
The survival of Salmonella on fresh ginger root (Zingiber officinale) during drying was examined using both a laboratory oven at 51 and 60°C with two different fan settings and a small commercially available food dehydrator. The survival of Salmonella in ground ginger stored at 25 and 37°C at 33% (low) and 97% (high) relative humidity (RH) was also examined. To inoculate ginger, a four-serovar cocktail of Salmonella was collected by harvesting agar lawn cells. For drying experiments, ginger slices (1 ± 0.5 mm thickness) were surface inoculated at a starting level of approximately 9 log CFU/g. Higher temperature (60°C) coupled with a slow fan speed (nonstringent condition) to promote a slower reduction in the water activity (aw) of the ginger resulted in a 3- to 4-log reduction in Salmonella populations in the first 4 to 6 h with an additional 2- to 3-log reduction by 24 h. Higher temperature with a higher fan speed (stringent condition) resulted in significantly less destruction of Salmonella throughout the 24-h period (P < 0.001). Survival appeared related to the rate of reduction in the aw. The aw also influenced Salmonella survival during storage of ground ginger. During storage at 97% RH, the maximum aw values were 0.85 at 25°C and 0.87 at 37°C; Salmonella was no longer detected after 25 and 5 days of storage, respectively, under these conditions. At 33% RH, the aw stabilized to approximately 0.35 at 25°C and 0.31 at 37°C. Salmonella levels remained relatively constant throughout the 365-day and 170-day storage periods for the respective temperatures. These results indicate a relationship between temperature and aw and the survival of Salmonella during both drying and storage of ginger.
Sigma-Aldrich
Potassium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium sulfate, suitable for plant cell culture
Sigma-Aldrich
Potassium sulfate, 99.99% trace metals basis
Sigma-Aldrich
Potassium sulfate, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Potassium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium sulfate, ACS reagent, ≥99.0%, powder or crystals
SAFC
Potassium sulfate
Sigma-Aldrich
Potassium sulfate, ACS reagent, ≥99.0%, powder