Merck
  • Variation of electronic transitions and reduction potentials of cerium(IV) complexes.
Variation of electronic transitions and reduction potentials of cerium(IV) complexes.
Dalton transactions (Cambridge, England : 2003) (2014-08-26)
Ursula J Williams, David Schneider, Walter L Dorfner, Cäcilia Maichle-Mössmer, Patrick J Carroll, Reiner Anwander, Eric J Schelter
ABSTRACT
The trivalent compound K[Ce[N(SiHMe2)2]4] was synthesized and oxidized, providing a convenient route to the reported cerium(IV) compound Ce[N(SiHMe2)2]4. Protonolysis reactions of Ce[N(SiHMe2)2]4 with tert-butanol, substituted benzyl alcohols, and 2,6-diphenylphenol yielded the neutral tetravalent compounds Ce(O(t)Bu)4(py)2, Ce2(OCH2C6R5)8(thf)2 (R = Me, F), and Ce(Odpp)4 (dpp = 2,6-(C6H5)2-C6H3). Spectroscopic and electrochemical characterization of the monometallic cerium(IV) silylamide, alkoxide, and aryloxide compounds revealed variable ligand-to-metal charge transfer transitions and metal-based reduction potentials. Computational bonding analyses were performed to complement the physical characterization of the complexes.
Sigma-Aldrich
Benzene-D6, 99.6 atom % D
Sigma-Aldrich
Benzene-D6, anhydrous, ≥99.6 atom % D
Sigma-Aldrich
Benzene-D6, "100%", 99.96 atom % D
Supelco
Pyridine, analytical standard
Supelco
Dichloromethane, Selectophore, ≥99.5%
Supelco
Toluene, analytical standard
Sigma-Aldrich
Benzene-D6, 99 atom % D
Sigma-Aldrich
tert-Butanol, anhydrous, ≥99.5%
Sigma-Aldrich
2,6-Diphenylphenol, 98%
Supelco
Pentane, analytical standard
Supelco
Hexane, analytical standard
Supelco
Dichloromethane, analytical standard
Supelco
tert-Butanol, analytical standard
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Ferrocene, 98%
Sigma-Aldrich
Benzene-D6, 99.6 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Pentane, ≥99% (GC)
Sigma-Aldrich
tert-Butanol, ACS reagent, ≥99.0%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
tert-Butanol, ≥99% (GC)
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
tert-Butanol, suitable for HPLC, ≥99.5%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%