Merck
  • In vitro antitrypanosomal and antiplasmodial activities of crude extracts and essential oils of Ocimum gratissimum Linn from Benin and influence of vegetative stage.
In vitro antitrypanosomal and antiplasmodial activities of crude extracts and essential oils of Ocimum gratissimum Linn from Benin and influence of vegetative stage.
Journal of ethnopharmacology (2014-07-25)
Bénédicta G H Kpadonou Kpoviessi, Salomé D S Kpoviessi, Eléonore Yayi Ladekan, Fernand Gbaguidi, Michel Frédérich, Mansourou Moudachirou, Joëlle Quetin-Leclercq, Georges C Accrombessi, Joanne Bero
ABSTRACT
Different parts of Ocimum gratissimum Linn are largely used in folk medicine for the treatment of many diseases, some of which related to parasitical infections as fevers and headaches. In order to validate their use and to clarify the plant part which possesses the best antiparasitic properties, we decided to evaluate the in vitro antiplasmodial and antitrypanosomal activities of essential oils and crude extracts from leaves, stems and seeds of Ocimum gratissimum as well as their cytotoxicity. The essential oils and ethanol crude extracts of leaves and stems of Ocimum gratissimum from Benin, were obtained in pre and full flowering stages. Seeds obtained only in full flowering stage, were also extracted. The oils were isolated by hydrodistillation and analyzed by GC/MS and GC/FID. Extracts and essential oils were tested in vitro against Trypanosoma brucei brucei and Plasmodium falciparum. Cytotoxicity was evaluated in vitro against Chinese Hamster Ovary (CHO) cells and the human non cancer fibroblast cell line (WI38) through MTT assay to evaluate the selectivity and toxicity was assessed against Artemia salina Leach. The essential oils and non-volatile crude extracts of Ocimum gratissimum were more active on Trypanosoma brucei brucei than on Plasmodium falciparum (3D7). This activity varies according to the vegetative stage (pre and full flowering) and the plant part (seeds, stems and leaves) extracted. The best growth inhibition of Trypanosoma brucei brucei was observed with ethanol crude extracts of leaves (IC50=1.66 ± 0.48 μg/mL) and seeds (IC50=1.29 ± 0.42 μg/mL) in full flowering stage with good selectivity (SI>10). The chemical composition of the essential oil from aerial parts (47 compounds), characterized by the presence as main constituents of p-cymene, thymol, γ-terpinene, β-myrcene and α-thujene, depends on the vegetative stage. The oil contained some minor compounds such as myrcene (IC50=2.24 ± 0.27μg/mL), citronellal (IC50=2.76 ± 1.55μg/mL), limonene (IC50=4.24 ± 2.27μg/mL), with good antitrypanosomal activities. These oils and crude extracts were not toxic against Artemia salina Leach and had a low cytotoxicity except leaves and seeds ethanol extracts obtained in full flowering which showed toxicity against CHO and WI38 cells. Our study shows that ethanol crude extracts of leaves and seeds of Ocimum gratissimum in full flowering stage can be a good source of antitrypanosomal agents. This is the first report about the relation between the plant part extracted, the vegetative stage of the plant, the antitrypanosomal and antiplasmodial activities and the cytotoxicity of essential oils and non-volatile extracts of Ocimum gratissimum from Benin.
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Supelco
Camphene, analytical standard
Sigma-Aldrich
Camphene, 95%
Sigma-Aldrich
Dimethyl ether, puriss., ≥99.9% (GC)
Sigma-Aldrich
Dimethyl ether, puriss., ≥99.9% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Artemisinin, 98%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl ether, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Camphene, primary reference standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Artemisinin, analytical standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Geraniol, ≥97%, FCC, FG
Sigma-Aldrich
(±)-Camphor, ≥95.5%
Sigma-Aldrich
Geraniol, natural, ≥97%, FG
Sigma-Aldrich
Geraniol, 98%