Merck
  • Probing the electrostatics of active site microenvironments along the catalytic cycle for Escherichia coli dihydrofolate reductase.
Probing the electrostatics of active site microenvironments along the catalytic cycle for Escherichia coli dihydrofolate reductase.
Journal of the American Chemical Society (2014-07-01)
C Tony Liu, Joshua P Layfield, Robert J Stewart, Jarrod B French, Philip Hanoian, John B Asbury, Sharon Hammes-Schiffer, Stephen J Benkovic
ABSTRACT
Electrostatic interactions play an important role in enzyme catalysis by guiding ligand binding and facilitating chemical reactions. These electrostatic interactions are modulated by conformational changes occurring over the catalytic cycle. Herein, the changes in active site electrostatic microenvironments are examined for all enzyme complexes along the catalytic cycle of Escherichia coli dihydrofolate reductase (ecDHFR) by incorporation of thiocyanate probes at two site-specific locations in the active site. The electrostatics and degree of hydration of the microenvironments surrounding the probes are investigated with spectroscopic techniques and mixed quantum mechanical/molecular mechanical (QM/MM) calculations. Changes in the electrostatic microenvironments along the catalytic environment lead to different nitrile (CN) vibrational stretching frequencies and (13)C NMR chemical shifts. These environmental changes arise from protein conformational rearrangements during catalysis. The QM/MM calculations reproduce the experimentally measured vibrational frequency shifts of the thiocyanate probes across the catalyzed hydride transfer step, which spans the closed and occluded conformations of the enzyme. Analysis of the molecular dynamics trajectories provides insight into the conformational changes occurring between these two states and the resulting changes in classical electrostatics and specific hydrogen-bonding interactions. The electric fields along the CN axes of the probes are decomposed into contributions from specific residues, ligands, and solvent molecules that make up the microenvironments around the probes. Moreover, calculation of the electric field along the hydride donor-acceptor axis, along with decomposition of this field into specific contributions, indicates that the cofactor and substrate, as well as the enzyme, impose a substantial electric field that facilitates hydride transfer. Overall, experimental and theoretical data provide evidence for significant electrostatic changes in the active site microenvironments due to conformational motion occurring over the catalytic cycle of ecDHFR.
Product Description
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Methotrexate for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, purum p.a., anhydrous, ≥98.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
2-Nitro-5-thiocyanatobenzoic acid, powder
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Folic acid, meets USP testing specifications
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
Folic acid, ≥97%
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Folic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Folic acid, Pharmaceutical Secondary Standard; Certified Reference Material