Merck
  • Fisetin inhibits high-glucose-induced vascular inflammation in vitro and in vivo.
Fisetin inhibits high-glucose-induced vascular inflammation in vitro and in vivo.
Inflammation research : official journal of the European Histamine Research Society ... [et al.] (2014-06-14)
Soyoung Kwak, Sae-Kwang Ku, Jong-Sup Bae
ABSTRACT
Fisetin, an active compound isolated from flowering plants in the family Fabaceae, was reported to have antiviral, neuroprotective, and anti-inflammatory effects. Vascular inflammatory processes have been suggested to play key roles in the initiation and progression of atherosclerosis, a major complication of diabetes mellitus. Thus, we determined the ability of fisetin to suppress vascular inflammatory processes induced by high glucose (HG) in primary human umbilical vein endothelial cells (HUVECs) and mice. The effects of fisetin on HG-induced vascular inflammation were determined by measuring vascular permeability, leukocyte adhesion and migration, cell adhesion molecule (CAM) expression levels, reactive oxygen species (ROS) formation, and nuclear factor (NF)-κB activation. HG markedly increased vascular permeability, monocyte adhesion, expressions of CAMs, formation of ROS, and activation of NF-κB. Remarkably, all of the observed vascular inflammatory effects induced by HG were inhibited by pretreatment with fisetin. Vascular inflammatory responses induced by HG are critical events underlying the development of diabetic complications; therefore, our results suggest that fisetin possesses significant therapeutic effects against diabetic complications and atherosclerosis.
Product Description
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture