Merck
  • Transporter-mediated uptake of UDP-glucuronic acid by human liver microsomes: assay conditions, kinetics, and inhibition.
Transporter-mediated uptake of UDP-glucuronic acid by human liver microsomes: assay conditions, kinetics, and inhibition.
Drug metabolism and disposition: the biological fate of chemicals (2014-11-09)
Andrew Rowland, Peter I Mackenzie, John O Miners
ABSTRACT
This study characterized the kinetics, variability, and factors that affect UDP-glucuronic acid (UDP-GlcUA) uptake by human liver microsomes (HLM). Biphasic kinetics were observed for UDP-GlcUA uptake by HLM. Uptake affinities (assessed as Kd) of the high- and low-affinity components differed by more than an order of magnitude (13 ± 6 vs. 374 ± 175 µM), but were comparable in terms of the maximal rate of uptake, with mean Vmax values differing less than 2.3-fold (56 ± 26 vs. 131 ± 35 pmol/min per mg). Variability in total intrinsic transporter activity (Uint) for microsomal UDP-GlcUA uptake across 12 livers was less than 4-fold. Experiments performed to optimize the conditions for microsomal UDP-GlcUA uptake demonstrated that both components were trans-stimulated by preloading (luminal addition) with an alternate UDP-sugar, and essentially abolished by the thiol-alkylating agent N-ethylmaleimide. Furthermore, interaction studies undertaken with a panel of drugs, alternate UDP-sugars, and glucuronide conjugates, at low (2.5 μM) and high (1000 μM) UDP-GlcUA concentrations, demonstrated that both components were inhibited to varying extents. Notably, the nucleoside analogs zidovudine, stavudine, lamivudine, and acyclovir inhibited both the high- and low- affinity components of microsomal UDP-GlcUA uptake by >45% at an inhibitor concentration of 100 μM. Taken together, these data demonstrate that human liver microsomal UDP-GlcUA uptake involves multiple protein-mediated components, and raises the possibility of impaired in vivo glucuronidation activity resulting from inhibition of UDP-GlcUA uptake into the endoplasmic reticulum membrane by drugs and other compounds.
Product Description
Aciclovir, European Pharmacopoeia (EP) Reference Standard
Hymecromone, European Pharmacopoeia (EP) Reference Standard
Lamivudine, European Pharmacopoeia (EP) Reference Standard
Lamivudine for system suitability 1, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Lamivudine, ≥98% (HPLC), powder
Propofol, European Pharmacopoeia (EP) Reference Standard
USP
Lamivudine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Uridine 5′-diphosphoglucose disodium salt, ≥98.0% (HPLC)
Sigma-Aldrich
4-Methylumbelliferone, ≥98%
Sigma-Aldrich
Acycloguanosine, ≥99% (HPLC), powder
Sigma-Aldrich
p-Acetamidophenyl β-D-glucuronide sodium salt, >98% (TLC)
Sigma-Aldrich
Uridine 5′-diphospho-N-acetylglucosamine sodium salt, ≥98%
Supelco
Lamivudine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acyclovir, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acyclovir, United States Pharmacopeia (USP) Reference Standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Propofol for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Zidovudine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2,6-Diisopropylphenol, 97%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
1-Hydroxypyrene, 98%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Phenolphthalein solution, 0.5 wt. % in ethanol: water (1:1)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Phenolphthalein, ACS reagent
Sigma-Aldrich
N-Ethylmaleimide, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
N-Ethylmaleimide, crystalline, ≥98% (HPLC)