Merck
  • Studies on the collision-induced dissociation of adipoR agonists after electrospray ionization and their implementation in sports drug testing.
Studies on the collision-induced dissociation of adipoR agonists after electrospray ionization and their implementation in sports drug testing.
Journal of mass spectrometry : JMS (2015-03-25)
Josef Dib, Nils Schlörer, Wilhelm Schänzer, Mario Thevis
ABSTRACT
AdipoR agonists are small, orally active molecules capable of mimicking the protein adiponectin, which represents an adipokine with antidiabetic and antiatherogenic effects. Two adiponectin receptors were reported in the literature referred to as adipoR1 and adipoR2. Activation of these receptors stimulates mitochondrial biogenesis and results in an improved oxidative metabolism (via adipoR1) and increased insulin sensitivity (via adipoR2). Hence, adipoR agonists are potentially performance enhancing substances and targets of proactive and preventive anti-doping measures. In this study, two adipoR agonists termed AdipoRon and 112254 as well as two isotopically labeled internal standards (ISTDs) were synthesized in three-step reactions. The products were fully characterized by nuclear magnetic resonance spectroscopy (NMR), mass spectrometry (MS) and density functional theory (DFT) computation. Collision-induced dissociation pathways following electrospray ionization were suggested based on the determined elemental compositions of product ions, comparison to product ions derived from labeled analogs (ISTDs), H/D-exchange experiments and the results of DFT calculations. The most abundant product ions were found at m/z 174, tentatively assigned to protonated 1-benzyl-1,2,3,4-tetrahydropyridine for AdipoRon, and m/z 207, suggested as protonated 1-(4-methoxybenzyl)piperazine, for 112254. Notably, the loss of the heterocyclic ring (i.e. piperazine and piperidine, respectively) in a supposedly intramolecular elimination reaction was observed in both cases. A qualitative determination of both AdipoR agonists in human plasma was established and fully validated for doping control purposes. Validation items such as recovery (86-89%), specificity, linearity, lower limit of detection (1 ng/ml), intraday (3-18%) and interday (5-16%) precision as well as ion suppression or enhancement were determined. Based on these findings adipoR agonists can be implemented in sports drug testing procedures.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
1-Hydroxybenzotriazole hydrate, wetted with not less than 20 wt. % water, 97% dry basis
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Triethylamine, ≥99.5%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
1-Hydroxybenzotriazole hydrate, ≥97.0% dry basis (T)
Supelco
Methanol, analytical standard
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
1-Hydroxybenzotriazole hydrate, wetted with not less than 14 wt. % water, 98% dry basis
Sigma-Aldrich
L-Alanine methyl ester hydrochloride, 99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
4-Amino-1-benzylpiperidine, 98%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder