Merck
  • Cecal succinate elevated by some dietary polyphenols may inhibit colon cancer cell proliferation and angiogenesis.
Cecal succinate elevated by some dietary polyphenols may inhibit colon cancer cell proliferation and angiogenesis.
Journal of agricultural and food chemistry (2014-05-27)
Tomoaki Haraguchi, Tomoko Kayashima, Yukako Okazaki, Junji Inoue, Shigeru Mineo, Kiminori Matsubara, Ei Sakaguchi, Noriyuki Yanaka, Norihisa Kato
ABSTRACT
This study demonstrated that 0.5% dietary rutin, ellagic acid, or curcumin markedly increased cecal succinate levels in rats fed a high-fat diet, whereas catechin, caffeic acid, and quercetin did not. Other organic acids were modestly or hardly affected by polyphenols. To clarify the effects of succinate levels increased by polyphenols, this study examined the effects of succinate on the growth and proliferation of colon cancer cells and angiogenesis. The growth and proliferation of HT29 human colon cancer cells and angiogenesis in an ex vivo model were significantly inhibited by succinate at a dose close to that in the cecum of rats fed polyphenols. Furthermore, succinate inhibited the migration of human umbilical vein endothelial cells. These findings suggest that the consumption of some polyphenols affects the health and diseases of the large intestine by elevating succinate.
Product Description
Supelco
Curcumin, analytical standard
Supelco
Caffeic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
Sodium succinate dibasic, purum, anhydrous, ≥98.0% (NT)
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
USP
Curcumin, United States Pharmacopeia (USP) Reference Standard
Curcumin, primary reference standard
Supelco
Curcumin, matrix substance for MALDI-MS, ≥99.5% (HPLC)
Sigma-Aldrich
Disodium succinate, ≥98%
Sigma-Aldrich
Vascular Endothelial Growth Factor human, VEGF, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Ellagic acid, ≥95% (HPLC), powder, from tree bark
Supelco
Ellagic acid, analytical standard
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Quercetin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
VEGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture