Merck
One-pot synthesis of antibacterial monomers with dual biocidal modes.
Journal of dentistry (2014-06-17)
Wei Zhang, Xiao-Juan Luo, Li-Na Niu, Si-Ying Liu, Wan-Chun Zhu, Jeevanie Epasinghe, Liang Chen, Guo-Hua Li, Cui Huang, Jing Mao, David H Pashley, Franklin R Tay
ABSTRACT
The present study reported a method for preparing a blend of antibacterial quaternary ammonium silanes and quaternary ammonium methacryloxy silane (QAMS) based on the sol-gel reaction between dimethyldiethoxy silane and two trialkoxysilanes, one with an antibacterial quaternary ammonium functionality and the other with a methacryloxy functionality. Reaction products of the sol-gel reaction were characterised by direct infusion mass spectrometry, FTIR and proton, carbon and silicon NMR. This blend of monomers was incorporated into an experimental universal adhesive for evaluation of antimicrobial activity against Streptococcus mutans biofilms, microtensile bond strength and cytotoxicty. Retention of quaternary ammonium species on polymerised adhesive, leaching of these species from the adhesive and the ability of resin-dentine interfaces to inhibit S. mutans biofilms were evaluated over a 3-month water-ageing period. The antibacterial adhesive version killed bacteria in S. mutans biofilms not only through the release of non-copolymerisable quaternary ammonium silane species (release-killing), but also via immobilised quaternary ammonium methacryloxy silane that are copolymerised with adhesive resin comonomers (contact-killing). Contact-killing was retained after water-ageing. The QAMS-containing universal adhesive has similar tensile bond strength as the control and two commercially available universal adhesives, when it was used for bonding to dentine in the etch-and-rinse mode and self-etching mode. Incorporation of the antimicrobial quaternary ammonium species blend did not adversely affect the cytotoxicity of the universal adhesive formulation. Instead of using quaternary ammonium dimethacrylates and nanosilver, an alternative bimodal antimicrobial strategy for formulating antimicrobial universal dentine adhesives is achieved using the one-pot sol-gel synthesis scheme. The QAMS containing universal dentine adhesives with dual antimicrobial activity is a promising material aimed at preventing second caries and prolonging the longevity of resin composite restorations.
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Sigma-Aldrich
Diethoxydimethylsilane, 97%
Sigma-Aldrich
Eugenol, natural, ≥98%, FG
Sigma-Aldrich
Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium chloride solution, 42 wt. % in methanol
Sigma-Aldrich
Eugenol, ≥98%, FCC, FG
Sigma-Aldrich
Cetyltrimethylammonium chloride solution, 25 wt. % in H2O
Supelco
Eugenol, PESTANAL®, analytical standard
Sigma-Aldrich
Diethoxydimethylsilane, purum, ≥97.0% (GC)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Hexadecyltrimethylammonium chloride, ≥98.0% (NT)
Supelco
Eugenol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Supelco
Eugenol, Pharmaceutical Secondary Standard; Certified Reference Material
Eugenol, European Pharmacopoeia (EP) Reference Standard
Supelco
Menadione (K3), analytical standard
Sigma-Aldrich
Fluorescein sodium salt, p.a., for source staining
Sigma-Aldrich
Fluorescein sodium salt, BioReagent, suitable for fluorescence
Sigma-Aldrich
Menadione, meets USP testing specifications
Sigma-Aldrich
Menadione, crystalline
Sigma-Aldrich
Fluorescein sodium salt, used as fluorescent tracer
Supelco
Menadione, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Fluorescein sodium salt, analytical standard
Menadione, European Pharmacopoeia (EP) Reference Standard
USP
Menadione, United States Pharmacopeia (USP) Reference Standard