Merck
  • Novel tubulin and tau neuroprotective fragments sharing structural similarities with the drug candidate NAP (Davuentide).
Novel tubulin and tau neuroprotective fragments sharing structural similarities with the drug candidate NAP (Davuentide).
Journal of Alzheimer's disease : JAD (2014-02-08)
Illana Gozes, Tal Iram, Evgenia Maryanovsky, Carmit Arviv, Liora Rozenberg, Yulie Schirer, Eliezer Giladi, Sharon Furman-Assaf
ABSTRACT
NAP (NAPVSIPQ, davunetide) is a microtubule stabilizing peptide drug candidate. Here, we set out to identify NAP-like peptides that provide neuroprotection and reduce tau pathology. NAP-like peptides were derived using publically available search engines, which identified sequence homologies in the microtubule subunit tubulin and in the microtubule associated protein, tau. NATLSIHQ (NAT) and STPTAIPQ were derived from tubulin, and TAPVPMPD (TAP) was derived from tau. All peptides provided neuroprotection against the Alzheimer's disease (AD) toxin, the amyloid-β 1-42 peptide, although NAT and TAP were much more potent than STPTAIPQ. NAT also protected astrocytes, while STPTAIPQ was active only at micromolar concentrations. Because NAT and TAP were much more potent than STPTAIPQ in neuroprotection, those peptides were also tested for inhibition of tau-like aggregation (the second protein hallmark pathology of AD). Both NAT and TAP inhibited tau-like aggregation, with NAT being active over a very broad concentration range. NAT also protected in vivo in a frontotemporal dementia transgenic mouse model (Tau-Tg), when tested at the age of ~10 months. Results showed significantly decreased levels of the NAP parent protein, activity-dependent neuroprotective protein in the cerebral cortex of the Tau-Tg which was increased back to normal levels by NAT treatment. This was coupled to protection of Brain-Body weight ratio in the compromised Tau-Tg. With AD being the major tauopathy and with tau taking part in frontotemporal dementia, novel NAP derivatives that reduce tauopathy and provide neuroprotection are of basic and clinical interest.
Sigma-Aldrich
Uridine, ≥99%
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Uridine, BioUltra, ≥99%
Sigma-Aldrich
Trizma® hydrochloride, BioXtra, pH 3.5-5.0 (0.5 M in H2O), ≥99.0% (titration)
Sigma-Aldrich
Trizma® hydrochloride, BioPerformance Certified, suitable for cell culture, ≥99.0% (titration)
Sigma-Aldrich
Trizma® hydrochloride, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Trizma® hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Sigma-Aldrich
Trizma® hydrochloride, reagent grade, ≥99.0% (titration), crystalline
Sigma-Aldrich
5-Fluoro-2′-deoxyuridine, thymidylate synthase inhibitor
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)