Merck
  • microRNA 376a regulates follicle assembly by targeting Pcna in fetal and neonatal mouse ovaries.
microRNA 376a regulates follicle assembly by targeting Pcna in fetal and neonatal mouse ovaries.
Reproduction (Cambridge, England) (2014-04-02)
Huan Zhang, Xiaohua Jiang, Yuanwei Zhang, Bo Xu, Juan Hua, Tieliang Ma, Wei Zheng, Rui Sun, Wei Shen, Howard J Cooke, Qiaomei Hao, Jie Qiao, Qinghua Shi
ABSTRACT
In mammals, the primordial follicle pool, providing all oocytes available to a female throughout her reproductive life, is established perinatally. Dysregulation of primordial follicle assembly results in female reproductive diseases, such as premature ovarian insufficiency and infertility. Female mice lacking Dicer1 (Dicer), a gene required for biogenesis of microRNAs, show abnormal morphology of follicles and infertility. However, the contribution of individual microRNAs to primordial follicle assembly remains largely unknown. Here, we report that microRNA 376a (miR-376a) regulates primordial follicle assembly by modulating the expression of proliferating cell nuclear antigen (Pcna), a gene we previously reported to regulate primordial follicle assembly by regulating oocyte apoptosis in mouse ovaries. miR-376a was shown to be negatively correlated with Pcna mRNA expression in fetal and neonatal mouse ovaries and to directly bind to Pcna mRNA 3' untranslated region. Cultured 18.5 days postcoitum mouse ovaries transfected with miR-376a exhibited decreased Pcna expression both in protein and mRNA levels. Moreover, miR-376a overexpression significantly increased primordial follicles and reduced apoptosis of oocytes, which was very similar to those in ovaries co-transfected with miR-376a and siRNAs targeting Pcna. Taken together, our results demonstrate that miR-376a regulates primordial follicle assembly by modulating the expression of Pcna. To our knowledge, this is the first microRNA-target mRNA pair that has been reported to regulate mammalian primordial follicle assembly and further our understanding of the regulation of primordial follicle assembly.
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, BioUltra, ≥99%
Supelco
Digoxigenin, analytical standard
Digoxigenin, European Pharmacopoeia (EP) Reference Standard
Trolamine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Triethanolamine, ≥99.0% (GC)
Sigma-Aldrich
Triethanolamine, puriss., meets analytical specification of NF, ≥99% (GC)
Sigma-Aldrich
Acetic anhydride, ReagentPlus®, ≥99%
Sigma-Aldrich
Triethanolamine, reagent grade, 98%
Sigma-Aldrich
Triethanolamine, puriss. p.a., ≥99% (GC)
Supelco
Triethanolamine, analytical standard
Sigma-Aldrich
Triethanolamine, BioUltra, ≥99.5% (GC)