Merck
Magnesium in disease prevention and overall health.
Advances in nutrition (Bethesda, Md.) (2013-05-16)
Stella Lucia Volpe
ABSTRACT
Magnesium is the fourth most abundant mineral and the second most abundant intracellular divalent cation and has been recognized as a cofactor for >300 metabolic reactions in the body. Some of the processes in which magnesium is a cofactor include, but are not limited to, protein synthesis, cellular energy production and storage, reproduction, DNA and RNA synthesis, and stabilizing mitochondrial membranes. Magnesium also plays a critical role in nerve transmission, cardiac excitability, neuromuscular conduction, muscular contraction, vasomotor tone, blood pressure, and glucose and insulin metabolism. Because of magnesium's many functions within the body, it plays a major role in disease prevention and overall health. Low levels of magnesium have been associated with a number of chronic diseases including migraine headaches, Alzheimer's disease, cerebrovascular accident (stroke), hypertension, cardiovascular disease, and type 2 diabetes mellitus. Good food sources of magnesium include unrefined (whole) grains, spinach, nuts, legumes, and white potatoes (tubers). This review presents recent research in the areas of magnesium and chronic disease, with the goal of emphasizing magnesium's role in disease prevention and overall health.
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Sigma-Aldrich
Magnesium, ribbon, ≥99.0% Mg basis
Sigma-Aldrich
Magnesium, powder, ≥99%
Sigma-Aldrich
Magnesium, rod, diam. 6 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Magnesium, turnings, reagent grade, 98%
Sigma-Aldrich
Magnesium, ReagentPlus®, ribbon, ≥99% trace metals basis
Sigma-Aldrich
Magnesium, grit, ≥99.0% (KT)
Sigma-Aldrich
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Sigma-Aldrich
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Sigma-Aldrich
Magnesium, 20-230 mesh, reagent grade, 98%
Sigma-Aldrich
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Sigma-Aldrich
Magnesium, turnings, 5-25 mm, 99.95% trace metals basis
Magnesium, wire reel, 0.1m, diameter 1.0mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 1.1mm, as drawn, 99.9+%
Magnesium, microfoil, 25x25mm, thinness 1.0μm, specific density 174μg/cm2, 6 micron aluminum permanent support, 99.9%
Magnesium, wire reel, 0.5m, diameter 0.125mm, as drawn, 99.9+%
Magnesium, wire reel, 2m, diameter 1.1mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 0.25mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 1.1mm, as drawn, 99.9+%
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Magnesium, microfoil, 50x50mm, thinness 1.0μm, specific density 174μg/cm2, 6 micron aluminum permanent support, 99.9%
Magnesium, wire reel, 2m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, microleaf, 50x50mm, thinness 1.0μm, specific density 174μg/cm2, removable support, 99.9%
Magnesium, wire reel, 2m, diameter 1.0mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 0.125mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 0.4mm, as drawn, 99.9+%
Magnesium, wire reel, 1m, diameter 0.4mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 0.25mm, as drawn, 99.9+%