Skip to Content
Merck
All Photos(1)

Key Documents

723312

Sigma-Aldrich

Magnesium

in a Sure/Seal bottle, turnings, 37.5 mmol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mg
CAS Number:
Molecular Weight:
24.31
Beilstein:
4948473
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

1 mmHg ( 621 °C)

form

turnings

autoignition temp.

950 °F

composition

, 37.5 mmol

reaction suitability

reagent type: reductant

resistivity

4.46 μΩ-cm, 20°C

bp

1090 °C (lit.)

mp

648 °C (lit.)

density

1.74 g/mL at 25 °C (lit.)

SMILES string

[Mg]

InChI

1S/Mg

InChI key

FYYHWMGAXLPEAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium is the most common mineral found in the human body. It is a cofactor for more than 300 enzymes and is known to regulate physiological functions like blood pressure, muscle contraction, and neuromuscular conduction.[1][2] It can be used in the preparation of many biomaterials with biomedical applications.[3] Magnesium is widely used in the organic synthesis as an organomagnesium compound viz Grignard reagent.[4]

Application

Magnesium can be used:
  • To prepare 1,3-bis[2-(3-n-dodecylthienyl)]azulene, applicable as an acid sensor material.[5]
  • As a catalyst in the cleavage of tetrahydrofuran to 4-chlorobutyl esters.[6]
  • To prepare copper-based composite materials with improved mechanical properties.[7]

Other Notes

Based on preparing 0.5 M solution of organomagnesium reagent and 40% headspace

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Pyr. Sol. 1 - Water-react 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Configuration-dependent optical properties and acid susceptibility of azulene compounds
Tang T, et al.
Journal of Material Chemistry C, 6(19), 5153-5160 (2018)
Magnesium and human health: perspectives and research directions
Al Alawi AM, et al.
International Journal of Endocrinology, 2018 (2018)
Development of a novel magnesium-copper based composite with improved mechanical properties
Hassan SF and Gupta M
Materials Research Bulletin, 37(2), 377-389 (2002)
The importance of magnesium in clinical healthcare
Schwalfenberg GK and Genuis SJ
Scientifica, 2017 (2017)
Use of magnesium (turnings) as a powerful catalyst for the O-acylative cleavage of tetrahydrofuran
Pasha MA and Manjula K
Synthetic Communications, 41(15), 2309-2314 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service