Merck
Biotin requirements for DNA damage prevention.
Mutation research (2011-08-30)
Janos Zempleni, Daniel Camara Teixeira, Toshinobu Kuroishi, Elizabeth L Cordonier, Scott Baier
ABSTRACT
Biotin serves as a covalently bound coenzyme in five human carboxylases; biotin is also attached to histones H2A, H3, and H4, although the abundance of biotinylated histones is low. Biotinylation of both carboxylases and histones is catalyzed by holocarboxylase synthetase. Human biotin requirements are unknown. Recommendations for adequate intake of biotin are based on the typical intake of biotin in an apparently healthy population, which is only a crude estimate of the true intake due to analytical problems. Importantly, intake recommendations do not take into account possible effects of biotin deficiency on impairing genome stability. Recent studies suggest that biotin deficiency causes de-repression of long terminal repeats, thereby causing genome instability. While it was originally proposed that these effects are caused by loss of biotinylated histones, more recent evidence suggests a more immediate role of holocarboxylase synthetase in forming multiprotein complexes in chromatin that are important for gene repression. Holocarboxylase synthetase appears to interact physically with the methyl-CpG-binding domain protein 2 and, perhaps, histone methyl transferases, thereby creating epigenetic synergies between biotinylation and methylation events. These observations might offer a mechanistic explanation for some of the birth defects seen in biotin-deficient animal models.
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Sigma-Aldrich
Biotin, meets USP testing specifications
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Biotin, European Pharmacopoeia (EP) Reference Standard
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Biotin, ≥99.0% (T)
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Biotin, United States Pharmacopeia (USP) Reference Standard
SAFC
BIOTIN