Merck
  • The rise of metal radionuclides in medical imaging: copper-64, zirconium-89 and yttrium-86.
The rise of metal radionuclides in medical imaging: copper-64, zirconium-89 and yttrium-86.
Future medicinal chemistry (2011-04-30)
Oluwatayo F Ikotun, Suzanne E Lapi
ABSTRACT
Positron emission tomography, with its high sensitivity and resolution, is growing rapidly as an imaging technology for the diagnosis of many disease states. The success of this modality is reliant on instrumentation and the development of effective and novel targeted probes. Initially, research in this area was focused on what we will define in this article as 'standard' PET isotopes (carbon-11, nitrogen-13, oxygen-15 and fluorine-18), but the short half-lives of these isotopes limit radiopharmaceutical development to those that probe rapid biological processes. To overcome these limitations, there has been a rise in nonstandard isotope probe development in recent years. This review focuses on the biological probes and processes that have been examined, in additiom to the preclinical and clinical findings with nonstandard radiometals: copper-64, zirconium-89, and yttrium-86.
Sigma-Aldrich
Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis
Sigma-Aldrich
Zirconium, sponge, ≥99% trace metals basis
Sigma-Aldrich
Zirconium, foil, thickness 0.1 mm, 99.98% trace metals basis
Zirconium, rod, 1000mm, diameter 6.0mm, centerless ground, 99.2%
Zirconium, rod, 100mm, diameter 1.5mm, 99.2%
Zirconium, rod, 500mm, diameter 3.0mm, centerless ground, 99.2%
Zirconium, wire reel, 1m, diameter 0.28mm, annealed, 99.2%
Zirconium, rod, 100mm, diameter 12.7mm, centerless ground, 99.2%
Zirconium, tube, 200mm, outside diameter 12.7mm, inside diameter 9.5mm, wall thickness 1.6mm, annealed, 99.2%
Zirconium, rod, 500mm, diameter 5.0mm, centerless ground, 99.2%
Zirconium, rod, 1000mm, diameter 9.5mm, centerless ground, 99.2%
Zirconium, tube, 1000mm, outside diameter 12.7mm, inside diameter 9.5mm, wall thickness 1.6mm, annealed, 99.2%
Zirconium, rod, 200mm, diameter 2.0mm, centerless ground, 99.2%
Zirconium, rod, 1000mm, diameter 5.0mm, centerless ground, 99.2%
Zirconium, rod, 100mm, diameter 19.0mm, centerless ground, 99.2%
Zirconium, foil, not light tested, 50x50mm, thickness 0.006mm, 99.2%
Zirconium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.2%
Zirconium, wire reel, 5m, diameter 0.25mm, as drawn, 99.2%
Zirconium, rod, 1000mm, diameter 1.5mm, 99.2%
Zirconium, tube, 200mm, outside diameter 6.35mm, inside diameter 4.55mm, wall thickness 0.9mm, annealed, 99.2%
Zirconium, rod, 500mm, diameter 9.5mm, centerless ground, 99.2%
Zirconium, rod, 100mm, diameter 3.0mm, centerless ground, 99.2%
Zirconium, wire reel, 5m, diameter 0.28mm, annealed, 99.2%
Zirconium, wire reel, 10m, diameter 0.25mm, as drawn, 99.2%
Zirconium, tube, 100mm, outside diameter 1.6mm, inside diameter 1.1mm, wall thickness 0.25mm, hard, 99.2%
Zirconium, rod, 200mm, diameter 5.0mm, centerless ground, 99.2%
Zirconium, rod, 500mm, diameter 2.0mm, centerless ground, 99.2%
Zirconium, tube, 500mm, outside diameter 1.6mm, inside diameter 1.1mm, wall thickness 0.25mm, hard, 99.2%
Zirconium, sponge, 50g, max. size 25mm, 99.2%
Zirconium, rod, 200mm, diameter 12.7mm, centerless ground, 99.2%