Merck
  • Postnatal hydrocortisone for preventing or treating bronchopulmonary dysplasia in preterm infants: a systematic review.
Postnatal hydrocortisone for preventing or treating bronchopulmonary dysplasia in preterm infants: a systematic review.
Neonatology (2010-02-13)
Lex W Doyle, Richard A Ehrenkranz, Henry L Halliday
ABSTRACT
Corticosteroids have been used widely after birth in preterm infants with respiratory failure; hydrocortisone may be preferable to other corticosteroids for this purpose. To determine if postnatal hydrocortisone is useful to prevent or treat bronchopulmonary dysplasia (BPD) in preterm infants. Randomised controlled trials (RCTs) of postnatal hydrocortisone therapy to prevent or treat BPD were sought. Data regarding clinical outcomes including mortality, BPD, death or BPD, complications during the primary hospitalisation, and long-term outcome were abstracted and analysed using RevMan 5. Eight RCTs enrolling a total of 880 participants were eligible. In all trials treatment was started in the first week of life; there were no trials of treatment started in infants who were chronically ventilator-dependent after the first week of life with established or evolving BPD. A meta-analysis of the available trials demonstrated little evidence for a direct effect of hydrocortisone on the rates of BPD, mortality, or the combined outcome of BPD or mortality. Hydrocortisone in the doses used in these eight studies had few beneficial or harmful effects; the notable exception was an increase in gastrointestinal perforation. Postnatal hydrocortisone in the doses and regimens used in the reported trials has few beneficial or harmful effects and cannot be recommended for prevention of BPD. There are no randomised trials to substantiate the use of hydrocortisone in chronically ventilator-dependent infants with established or evolving BPD.
Product Description
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cortisol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard