Merck
  • Derivatization and chromatographic behavior of the o-phthaldialdehyde amino acid derivatives obtained with various SH-group-containing additives.
Derivatization and chromatographic behavior of the o-phthaldialdehyde amino acid derivatives obtained with various SH-group-containing additives.
Journal of chromatography. A (2001-05-18)
I Molnár-Perl
ABSTRACT
An overview is presented of HPLC methods currently in use to determine amino acids as their o-phthaldialdyde derivatives in the presence of various SH-group-containing additives. Crucial points that proved to influence the stability of the amino acid OPA derivatives have been discussed in detail: (i) the mol ratios of the OPA-SH-group-containing additive amino acid; (ii) the preparation and storage conditions of the OPA reagents; (iii) the optimum pH conditions for the interactions and elutions; (iv) the behavior of the, believed to be, less stable amino acids, such as glycine, beta-alanine, gamma-aminobutyric acid, histidine, ornithine and lysine.
Sigma-Aldrich
Phthaldialdehyde, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder or crystals
Sigma-Aldrich
Phthaldialdehyde, ≥97% (HPLC), powder or crystals
Sigma-Aldrich
Phthaldialdehyde, ≥99.0% (HPLC)
Sigma-Aldrich
Phthaldialdehyde Reagent, Solution Complete
Sigma-Aldrich
Phthaldialdehyde Reagent, Solution Incomplete