Skip to Content
Merck
All Photos(1)

Key Documents

P7914

Sigma-Aldrich

Phthaldialdehyde Reagent

Solution Incomplete

Synonym(s):

OPA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

Quality Level

storage temp.

2-8°C

SMILES string

[H]C(=O)c1ccccc1C([H])=O

InChI

1S/C8H6O2/c9-5-7-3-1-2-4-8(7)6-10/h1-6H

InChI key

ZWLUXSQADUDCSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Phthaldialdehyde Reagent has been used for the derivatization plasma samples prior to high performance liquid chromatography.
Precolumn derivatization reagent for primary amines and amino acids. The fluorescent derivative can be detected by reverse-phase HPLC. The reaction requires OPA, primary amine and a sulfhydryl. In the presence of excess sulfhydryl, amines can be quantitated. In the presence of excess amine, sulfhydryls can be quantitated.
Useful for investigators requiring an alternate sulfhydryl moiety. Requires addition of desired sulfhydryl compound for measuring amines. Requires addition of amine to measure sulfhydryls.

Linkage

Same as P 0532, but contains no 2-mercaptoethanol.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Low-dose tryptophan depletion in recovered depressed patients induces changes in cognitive processing without depressive symptoms
Hayward G, et al.
Biological Psychiatry, 57(5), 517-524 (2005)
Gianni Pala et al.
Expert review of clinical immunology, 9(3), 227-234 (2013-03-01)
This study summarizes and reviews the available health information on ortho-phthalaldehyde (OPA), a recently introduced and widespread disinfectant for heat-sensitive medical equipment, particularly focusing on its possible immunological effects in the healthcare setting. OPA properties derived from laboratory and clinical
Jong-Suk Song et al.
Investigative ophthalmology & visual science, 53(10), 6164-6170 (2012-08-18)
We evaluated whether viscoelastics have protective effects on the corneal endothelial cell damage in a toxic anterior segment syndrome (TASS) animal model depending on the types of viscoelastics. A TASS animal model was established with an injection of 0.1 mL
Michal Douša et al.
Journal of pharmaceutical and biomedical analysis, 66, 359-364 (2012-04-06)
A rapid procedure based on direct extraction and HILIC separation of aliskiren (ALI) degradation product - 3-amino-2,2-dimethylpropanamide (ADPA) with fluorescence detection has been developed. The formation of ADPA from ALI under different conditions was studied. The evaluation of HILIC method
Stella E Hines et al.
Current opinion in allergy and clinical immunology, 12(2), 102-110 (2012-02-07)
Lymphocyte proliferation testing (LPT) is used in diagnosing occupationally acquired delayed-type hypersensitivity. It has been used in beryllium-health effects, and its role is expanding in metal allergy. It may find application in diagnosis of other sensitizers. Use of the beryllium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service