Merck
  • Surface enhanced fluorescence from silver film substrate decorated with nanohole arrays.
Surface enhanced fluorescence from silver film substrate decorated with nanohole arrays.
Journal of nanoscience and nanotechnology (2014-04-18)
Yu Sun, Xiao Qing Yan, Yabing Du, Zhenglong Zhang, Yiping Huo, Fei Gao, Hairong Zheng
ABSTRACT
The fluorescence enhancement effect of Rh6G molecules deposited on the silver film substrate decorated with nanohole arrays was investigated in this paper. The prepared substrate, decorated with nanohole arrays, was fabricated with the deposition of silver films onto the anodic aluminum oxide templates through magnetron sputtering method. Compared with the conventional continuous silver film substrate, the prepared substrate shows better enhanced effect. Particularly, the fluorescence enhancement factor has a relationship with the size and period of the nanohole arrays. The experimental observations were analyzed with local surface plasmon resonance model. The results of current work highlight the importance of strong electromagnetic coupling effect in surface enhanced fluorescence.
Sigma-Aldrich
Silver, conductive paste
Sigma-Aldrich
Silver, colloidal, 65-75% Ag basis
Sigma-Aldrich
Silver, wire, diam. 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, nanopowder, <150 nm particle size, 99% trace metals basis
Sigma-Aldrich
Silver, shot, 1-3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, powder, <250 μm, 99.99% trace metals basis
Sigma-Aldrich
Silver, wool, diam. 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Silver, foil, 10mm disks, thickness 0.5mm, as rolled, 99.99+%
Silver, wire reel, 1m, diameter 0.025mm, as drawn, 99.99%
Silver, wire reel, 20m, diameter 0.125mm, annealed, 99.99%
Silver, foil, 50x50mm, thickness 0.25mm, as rolled, 99.95+%
Silver, wire reel, 1m, diameter 0.050mm, gold plated, 99.99%
Silver, foil, 0.5m coil, thickness 0.15mm, as rolled, 99.95+%
Silver, foil, 10mm disks, thickness 0.008mm, 99.95+%
Silver, foil, 10mm disks, thickness 1.0mm, as rolled, 99.99+%
Silver, foil, 10mm disks, thickness 0.0125mm, 99.95+%
Silver, wire reel, 0.5m, diameter 0.25mm, as drawn, 99.997%
Silver, foil, 10mm disks, thickness 0.05mm, as rolled, 99.95+%
Silver, foil, 10mm disks, thickness 0.025mm, as rolled, 99.95+%
Silver, foil, 15mm disks, thickness 0.003mm, 99.9%
Silver, foil, 10m coil, thickness 0.0125mm, coil width 9mm, 99.95+%
Silver, foil, 10mm disks, thickness 0.02mm, 99.97%
Silver, foil, 10mm disks, thickness 0.125mm, as rolled, 99.99+%