Merck
  • Easily controllable synthesis of alpha-MoO3 nanobelts and MoO2 microaxletrees through one-pot hydrothermal route.
Easily controllable synthesis of alpha-MoO3 nanobelts and MoO2 microaxletrees through one-pot hydrothermal route.
Journal of nanoscience and nanotechnology (2014-04-18)
Hanmei Hu, Junchan Xu, Chonghai Deng, Xinqing Ge
ABSTRACT
A mild one-pot hydrothermal route has been successfully designed to controllably prepare orthorhombic alpha-MoO3 nanobelts and monoclinic MoO2 microaxletrees respectively by adjusting the dosage of (NH4)6M07O24 x 4H2O (AHM). The products are characterized by X-ray diffraction (XRD), field-emission scanning electron microscopy (FESEM), transmission electron microscopy (TEM) and UV-visible absorption spectrum. The as-prepared alpha-MoO3 nanobelts, with widths of 100-400 nm and lengths up to 30-40 microm, grow along [001] direction. The as-obtained MoO2 microaxletrees are assembled by countless nanolaths with the thickness of 80-150 nm. The chemical reaction processes for the formation of MoO(x) (MoO3 and MoO2) phases are investigated based on the experimental phenomena. The possible growth mechanisms are also discussed. The band gap energies (E(g)) of the obtained alpha-MoO3 nanobelts and MoO2 microaxletrees are calculated to be 2.90 and 3.72 eV, respectively. This work exhibits an effective approach in the selectively controlled synthesis of MoO(x) (x = 2, 3) nanomaterials via one-step hydrothermal strategy.
Product Description
Supelco
Water, for TOC analysis
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Molybdenum(VI) oxide, nanopowder, 100 nm (TEM), 99.5% trace metals basis
Molybdenum, rod, 100mm, diameter 8.0mm, centerless ground, 99.9%
Molybdenum, tube, 300mm, outside diameter 0.5mm, inside diameter 0.38mm, wall thickness 0.06mm, 99.9%
Molybdenum, rod, 500mm, diameter 1.5mm, centerless ground, 99.9%
Molybdenum, tube, 100mm, outside diameter 2.0mm, inside diameter 1.3mm, wall thickness 0.35mm, 99.9%
Molybdenum, tube, 50mm, outside diameter 6.4mm, inside diameter 4.6mm, wall thickness 0.9mm, 99.9%
Molybdenum, rod, 100mm, diameter 25mm, centerless ground, 99.9%
Molybdenum, rod, 500mm, diameter 2.0mm, centerless ground, 99.9%
Molybdenum, tube, 500mm, outside diameter 2.0mm, inside diameter 1.3mm, wall thickness 0.35mm, 99.9%
Molybdenum, rod, 500mm, diameter 15.0mm, centerless ground, 99.9%
Molybdenum, rod, 50mm, diameter 5.0mm, 99.98%
Molybdenum, rod, 100mm, diameter 6.0mm, centerless ground, 99.9%
Molybdenum, rod, 200mm, diameter 10.0mm, centerless ground, 99.9%
Molybdenum, rod, 200mm, diameter 25mm, centerless ground, 99.9%
Molybdenum, tube, 200mm, outside diameter 1.0mm, inside diameter 0.7mm, wall thickness 0.15mm, 99.9%
Molybdenum, rod, 100mm, diameter 15.0mm, centerless ground, 99.9%
Molybdenum, tube, 1000mm, outside diameter 1.0mm, inside diameter 0.7mm, wall thickness 0.15mm, 99.9%
Molybdenum, foil, 8mm disks, thickness 0.20mm, annealed, 99.9%
Molybdenum, foil, 8mm disks, thickness 0.0125mm, 99.9%
Molybdenum, foil, 8mm disks, thickness 0.15mm, annealed, 99.9%
Molybdenum, foil, light tested, 100x100mm, thickness 0.01mm, 99.9%
Molybdenum, foil, 8mm disks, thickness 0.1mm, annealed, 99.9%
Molybdenum, foil, 6mm disks, thickness 0.5mm, annealed, 99.9%
Molybdenum, tube, 500mm, outside diameter 3.15mm, inside diameter 2.39mm, wall thickness 0.38mm, 99.9%
Molybdenum, tube, 100mm, outside diameter 6.0mm, inside diameter 5.6mm, wall thickness 0.20mm, 99.9%
Molybdenum, rod, 50mm, diameter 20.0mm, centerless ground, 99.9%