Skip to Content
Merck
All Photos(1)

Key Documents

326496

Sigma-Aldrich

Gold

foil, thickness 0.127 mm, 99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Au
CAS Number:
Molecular Weight:
196.97
EC Number:
MDL number:
UNSPSC Code:
12141717
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.99% trace metals basis

form

foil

resistivity

2.05 μΩ-cm, 0°C

thickness

0.127 mm

bp

2808 °C (lit.)

mp

1063 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Related Categories

Application

Gold-foil can be used to fabricate carbide-derived nanoporous carbon based polymeric actuators. Gold-foil modified actuators show high frequency response and low operating voltage.

TiO2 coated gold foils can used to enhance the electrocatalytic oxidation of glucose.

Gold foil can be used as a reference electrode for the electro-reduction of carbon dioxide.

Quantity

1.5 g = 25 × 25 mm; 6 g = 50 × 50 mm

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Zhengxia Liu et al.
Journal of biomedical nanotechnology, 9(6), 1017-1028 (2013-07-19)
Ischemic stroke is a leading cause of death and disability, and the treatment options are limited. Recent studies demonstrated that the promising applications of gold nanoparticles (Au-NPs) as intrinsic therapeutics; however, little is known about the effect of Au-NPs on
Young Joo Choi et al.
Journal of nanoscience and nanotechnology, 13(6), 4437-4445 (2013-07-19)
Gold nanorods (Au NRs) that absorb near-infrared (NIR) light have great potential in the field of nanomedicine. Photothermal therapy (PTT), a very attractive cancer therapy in nanomedicine, combines nanomaterials and light. The aim of this study was to elucidate the
Tae-Sik Cho et al.
Journal of nanoscience and nanotechnology, 13(5), 3711-3714 (2013-07-19)
The crystallization of Au/glass ultrathin films for surface plasmon resonance (SPR) biosensor has been studied using synchrotron X-ray scattering and field emission scanning electron microscope. In films thinner than 30 nm, crystallized Au grains with [111] preferred orientation were formed
Matthieu B Miller et al.
Journal of the Air & Waste Management Association (1995), 63(6), 681-693 (2013-07-19)
Industrial gold mining is a significant source of mercury (Hg) emission to the atmosphere. To investigate ways to reduce these emissions, reclamation and dust and mercury control methods used at open pit gold mining operations in Nevada were studied in
Nidhi Gupta et al.
Journal of nanoscience and nanotechnology, 13(7), 4917-4924 (2013-08-02)
The preparation and photocatalytic reductive ability of a new spongy flower or cotton bud like amorphous CdS microstructures (size = 200-300 nm) having intermediate crystal phase between cubic and hexagonal structure and possesses much larger surface area ca. 119 m2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service