Merck
  • An overview of Y-Family DNA polymerases and a case study of human DNA polymerase η.
An overview of Y-Family DNA polymerases and a case study of human DNA polymerase η.
Biochemistry (2014-04-11)
Wei Yang
ABSTRACT
Y-Family DNA polymerases specialize in translesion synthesis, bypassing damaged bases that would otherwise block the normal progression of replication forks. Y-Family polymerases have unique structural features that allow them to bind damaged DNA and use a modified template base to direct nucleotide incorporation. Each Y-Family polymerase is unique and has different preferences for lesions to bypass and for dNTPs to incorporate. Y-Family polymerases are also characterized by a low catalytic efficiency, a low processivity, and a low fidelity on normal DNA. Recruitment of these specialized polymerases to replication forks is therefore regulated. The catalytic center of the Y-Family polymerases is highly conserved and homologous to that of high-fidelity and high-processivity DNA replicases. In this review, structural differences between Y-Family and A- and B-Family polymerases are compared and correlated with their functional differences. A time-resolved X-ray crystallographic study of the DNA synthesis reaction catalyzed by the Y-Family DNA polymerase human polymerase η revealed transient elements that led to the nucleotidyl-transfer reaction.
Sigma-Aldrich
Magnesium, ReagentPlus®, ribbon, ≥99% trace metals basis
Sigma-Aldrich
Magnesium, rod, diam. 6 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Sigma-Aldrich
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Sigma-Aldrich
Magnesium, powder, ≥99%
Sigma-Aldrich
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Sigma-Aldrich
Magnesium, 20-230 mesh, reagent grade, 98%
Sigma-Aldrich
Magnesium, turnings, reagent grade, 98%
Sigma-Aldrich
Magnesium preparation, Highly Reactive Rieke® Metal, suspension, 2.5 g in THF
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer containing MgCl2
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer without MgCl2
Sigma-Aldrich
Magnesium, ribbon, ≥99.0% Mg basis
Sigma-Aldrich
Magnesium, grit, ≥99.0% (KT)
Sigma-Aldrich
Magnesium, turnings, 5-25 mm, 99.95% trace metals basis
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Magnesium, microleaf, 50x50mm, thinness 1.0μm, specific density 174μg/cm2, removable support, 99.9%
Magnesium, wire reel, 2m, diameter 1.0mm, as drawn, 99.9+%
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Magnesium, wire reel, 1m, diameter 1.0mm, as drawn, 99.9+%
Magnesium, wire reel, 2m, diameter 0.125mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 1.1mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 1.1mm, as drawn, 99.9+%
Magnesium, microfoil, 50x50mm, thinness 1.0μm, specific density 174μg/cm2, 6 micron aluminum permanent support, 99.9%
Magnesium, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.9+%
Magnesium, wire reel, 2m, diameter 1.1mm, as drawn, 99.9+%
Magnesium, wire reel, 1m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 0.25mm, as drawn, 99.9+%
Magnesium, wire reel, 2m, diameter 0.5mm, as drawn, 99.9+%