Sigma-Aldrich
Nickel, wire, diam. 0.25 mm, ≥99.9%
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 4200, slurry, in H2O, active catalyst
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 3202, slurry, in H2O, active catalyst
Sigma-Aldrich
Nickel, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.25 mm, 99.995% trace metals basis
Sigma-Aldrich
Nickel, powder, <1 μm, 99.8% trace metals basis
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2800, slurry, in H2O, active catalyst
Sigma-Aldrich
Nickel, foil, thickness 0.125 mm, ≥99.9%
Sigma-Aldrich
Nickel, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
Nickel, foil, 25mm disks, thickness 0.25mm, as rolled, 100%
Nickel, microfoil, disks, 10mm, thinness 1.0μm, specific density 895.8μg/cm2, permanent mylar 3.5μm support, 99.99+%
Nickel, foil, not light tested, 100x100mm, thickness 0.0125mm, 99.9%
Nickel, foil, 2m coil, thickness 0.0075mm, 99.9%
Nickel, foil, not light tested, 25x25mm, thickness 0.002mm, 99.95%
Nickel, foil, 25x25mm, thickness 1.0mm, as rolled, 99.99+%
Nickel, foil, light tested, 25x25mm, thickness 0.025mm, as rolled, 100%
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2400, slurry, in H2O, active catalyst
Nickel, foil, light tested, 50x50mm, thickness 0.0075mm, 99.95%
Nickel, foil, 2m coil, thickness 0.20mm, annealed, 99%
Nickel, foil, 150x150mm, thickness 0.3mm, annealed, 99%
Nickel, mesh, 100x250mm, nominal aperture 0.73mm, wire diameter 0.25mm, 26x26 wires/inch, open area 55%, plain weave mesh, 99%
Nickel, foil, 10mm disks, thickness 0.02mm, 99.9%
Nickel, foil, 0.5m coil, thickness 0.125mm, as rolled, 99.99+%
Nickel, foil, 10m coil, thickness 0.20mm, coil width 1mm, annealed, 99%
Nickel, foil, 4mm disks, thickness 0.0125mm, 99.9%
Nickel, microfoil, disks, 10mm, thinness 0.25μm, specific density 222.55μg/cm2, permanent mylar 3.5μm support, 99.99+%
Nickel, wire reel, 0.2m, diameter 0.4mm, as drawn, 99.99+%
Nickel, foil, 10mm disks, thickness 0.038mm, 99.9%
Nickel, foil, 2m coil, thickness 0.3mm, coil width 1mm, annealed, 99%