Skip to Content
Merck
All Photos(1)

Documents

510068

Sigma-Aldrich

Raney®-Nickel

W.R. Grace and Co. Raney® 3202, slurry, in H2O, active catalyst

Synonym(s):

Nickel sponge

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AlNi
CAS Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

slurry

reaction suitability

core: nickel
reagent type: catalyst

manufacturer/tradename

W.R. Grace and Co. Raney® 3202

concentration

in H2O

SMILES string

[Al].[Ni]

InChI

1S/Al.Ni

InChI key

NPXOKRUENSOPAO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Useful as a selective reducing agent for hydrogenation of organic compounds

Legal Information

Raney is a registered trademark of W.R. Grace & Co.-Conn.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Pyr. Liq. 1 - Skin Sens. 1 - STOT RE 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nitrile reduction in the presence of Boc-protected amino groups by catalytic hydrogenation over palladium-activated Raney-nickel.
B Klenke et al.
The Journal of organic chemistry, 66(7), 2480-2483 (2001-04-03)
Tsukinoki, T.; Kanda, T.
Tetrahedron Letters, 41, 5865-5865 (2000)
Barrero, A. F.; Alvarez-Manzaneda, E. J. et al.
Synthesis, 197-197 (2000)
Toth, M.; Somsak, L.,
Tetrahedron Letters, 42, 2723-2723 (2001)
S M Marchan et al.
The European journal of prosthodontics and restorative dentistry, 21(1), 24-28 (2013-05-21)
Posterior resin bonded cast metal restorations (adhesive onlays) were used in a variety of clinical scenarios including: management of tooth wear and cracked tooth, as retainers for fixed bridge work, for correction of the occlusal plane and in providing cuspal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service