Merck
In-vivo force decay of nickel-titanium closed-coil springs.
American journal of orthodontics and dentofacial orthopedics : official publication of the American Association of Orthodontists, its constituent societies, and the American Board of Orthodontics (2014-04-08)
Crystal Cox, Tung Nguyen, Lorne Koroluk, Ching-Chang Ko
ABSTRACT
Nickel-titanium closed-coil springs are purported to deliver constant forces over extended ranges of activation and working times. In-vivo studies supporting this claim are limited. The objective of this study was to evaluate changes in force-decay properties of nickel-titanium closed-coil springs after clinical use. Pseudoelastic force-deflection curves for 30 nickel-titanium coil springs (used intraorally) and 15 matched laboratory control springs (simulated intraoral conditions: artificial saliva, 37°C) were tested before and after retrieval via dynamic mechanical analysis and a testing machine, respectively, to evaluate the amounts of force-loss and hysteresis change after 4, 8, or 12 weeks of working time (n = 10 per group). The effects of the oral environment and clinical use on force properties were evaluated by comparing in-vivo and in-vitro data. The springs studied showed a statistically significant decrease in force (approximately 12%) after 4 weeks of clinical use (P <0.01), with a further significant decrease (approximately 7%) from 4 to 8 weeks (P = 0.03), and force levels appearing to remain steady thereafter. Clinical space closure at an average rate of 0.91 mm per month was still observed despite this decrease in force. In-vivo and in-vitro force-loss data were not statistically different. Nickel-titanium closed-coil springs do not deliver constant forces when used intraorally, but they still allow for space-closure rates of approximately 1 mm per month.
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Nickel, foil, 50mm disks, thickness 0.15mm, annealed, 99%
Nickel, foil, 5m coil, thickness 0.05mm, annealed, 99%
Nickel, foil, 50mm disks, thickness 0.0125mm, 99.99+%
Nickel, foil, 50mm disks, thickness 0.0075mm, 99.9%
Nickel, foil, 5m coil, thickness 0.05mm, coil width 2mm, annealed, 99%
Nickel, foil, 4mm disks, thickness 0.5mm, annealed, 99.98%
Nickel, foil, 50mm disks, thickness 0.125mm, as rolled, 99.99+%
Nickel, foil, 4mm disks, thickness 0.25mm, annealed, 99%
Nickel, foil, 50mm disks, thickness 0.03mm, 99.9%
Nickel, foil, 6mm disks, thickness 0.025mm, as rolled, 99.99+%
Nickel, foil, 6mm disks, thickness 0.005mm, as rolled, 99.9%
Nickel, foil, 6mm disks, thickness 0.125mm, annealed, 99.98%
Nickel, foil, 6mm disks, thickness 0.007mm, 99.95%
Nickel, foil, 6mm disks, thickness 0.02mm, 99.9%
Nickel, foil, 8mm disks, thickness 0.0125mm, 99.99+%
Nickel, foil, 8mm disks, thickness 0.05mm, 99.9%
Nickel, foil, 6mm disks, thickness 0.3mm, as rolled, 100%
Nickel, foil, 8mm disks, thickness 0.03mm, 99.9%
Nickel, foil, 6mm disks, thickness 0.0125mm, 99.9%
Nickel, broken cathode squares, 200g, max. size 50mm, 99.8%
Nickel, foil, 6mm disks, thickness 0.0125mm, annealed, 99%
Nickel, foil, 8mm disks, thickness 0.125mm, annealed, 99.98%
Nickel, foil, 6mm disks, thickness 0.038mm, 99.9%
Nickel, foil, 8mm disks, thickness 0.01mm, 99.95%