Merck
  • Modelling uptake and toxicity of nickel in solution to Enchytraeus crypticus with biotic ligand model theory.
Modelling uptake and toxicity of nickel in solution to Enchytraeus crypticus with biotic ligand model theory.
Environmental pollution (Barking, Essex : 1987) (2014-02-18)
Erkai He, Hao Qiu, Cornelis A M Van Gestel
ABSTRACT
Protons and other cations may inhibit metal uptake and alleviate metal toxicity in aquatic organisms, but less is known about these interactions in soil organisms. The present study investigated the influence of solution chemistry on uptake and toxicity of Ni in Enchytraeus crypticus after 14 days exposure. Ca(2+), Mg(2+) and Na(+) were found to exert significant effects on both uptake and toxicity of Ni. An extended Langmuir model, which incorporated cation competition effects, well predicted Ni uptake. The LC50{Ni(2+)} predicted by a developed Biotic Ligand Model matched well with observed values. These suggest that cation competition needs to be taken into account when modelling uptake and effects. The binding constants of Ni(2+), Mg(2+) and Na(+) on the uptake and toxic action sites were similar, but for Ca(2+) they differed. This indicates that the effect of Ca(2+) on Ni(2+) toxicity cannot simply be explained by the competition for entry into organism.
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Nickel, rod, 200mm, diameter 40mm, 99%
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Nickel, wire reel, 5m, diameter 0.5mm, as drawn, 99%
Nickel, wire reel, 2m, diameter 0.75mm, as drawn, 99.99+%
Nickel, wire reel, 500m, diameter 0.125mm, hard, 99.98%
Nickel, foil, 6mm disks, thickness 0.007mm, 99.95%
Nickel, wire reel, 5m, diameter 0.25mm, as drawn, 99.99+%
Nickel, wire reel, 5m, diameter 0.5mm, hard, 99.98%
Nickel, wire reel, 5m, diameter 0.05mm, as drawn, 99.99+%
Nickel, foil, 6mm disks, thickness 0.015mm, 99.9%
Nickel, foil, 6mm disks, thickness 0.05mm, annealed, 99%
Nickel, wire reel, 500m, diameter 0.25mm, hard, 99.98%
Nickel, foil, not light tested, 100x100mm, thickness 0.002mm, 99.95%
Nickel, foil, not light tested, 25x25mm, thickness 0.001mm, 99.95%
Nickel, foil, 150x300mm, thickness 0.25mm, annealed, 99.98%
Nickel, pellets, 50g, max. size 5mm, 99.99+%
Nickel, wire reel, 0.5m, diameter 0.05mm, as drawn, 99.99+%
Nickel, foil, 4mm disks, thickness 0.02mm, 99.9%
Nickel, foil, 0.5m coil, thickness 0.038mm, 99.9%
Nickel, foil, 0.2m coil, thickness 0.004mm, hard, 99%
Nickel, foil, 150x300mm, thickness 0.5mm, annealed, 99.98%
Nickel, foil, 10mm disks, thickness 0.0125mm, 99.9%
Nickel, foil, 0.5m coil, thickness 0.15mm, coil width 12mm, annealed, 99%
Nickel, foil, 50x50mm, thickness 1.0mm, annealed, 99%
Nickel, foil, 25x25mm, thickness 3.2mm, as rolled, 99.99+%