Merck
Intakes of potassium, magnesium, and calcium and risk of stroke.
Stroke (2014-02-13)
Ivonne Sluijs, Sebastien Czernichow, Joline W J Beulens, Jolanda M A Boer, Yvonne T van der Schouw, W Monique M Verschuren, Diederick E Grobbee
ABSTRACT
We aimed to investigate the associations of dietary and total potassium, magnesium, and calcium intakes with stroke occurrence. A prospective cohort study was conducted among 36 094 participants aged 21 to 70 years. Dietary intake was assessed with a food frequency questionnaire. During 12 years of follow-up, 631 strokes occurred. After adjustment for confounders, magnesium intake was associated with reduced stroke risk (hazard ratio [95% confidence interval] per 100 mg/d, 0.80 [0.67-0.97] dietary magnesium; 0.78 [0.65-0.93] total magnesium). Potassium and calcium intakes were not associated with stroke. This study supports an association between high magnesium intake and a reduced stroke risk.
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Sigma-Aldrich
Magnesium, ribbon, ≥99.0% Mg basis
Sigma-Aldrich
Magnesium, powder, ≥99%
Sigma-Aldrich
Magnesium, rod, diam. 6 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Magnesium, turnings, reagent grade, 98%
Sigma-Aldrich
Magnesium, ReagentPlus®, ribbon, ≥99% trace metals basis
Sigma-Aldrich
Magnesium, grit, ≥99.0% (KT)
Sigma-Aldrich
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Sigma-Aldrich
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Sigma-Aldrich
Magnesium, 20-230 mesh, reagent grade, 98%
Sigma-Aldrich
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Sigma-Aldrich
Magnesium, turnings, 5-25 mm, 99.95% trace metals basis
Magnesium, wire reel, 0.1m, diameter 1.0mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 1.1mm, as drawn, 99.9+%
Magnesium, microfoil, 25x25mm, thinness 1.0μm, specific density 174μg/cm2, 6 micron aluminum permanent support, 99.9%
Magnesium, wire reel, 0.5m, diameter 0.125mm, as drawn, 99.9+%
Magnesium, wire reel, 2m, diameter 1.1mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 0.25mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 1.1mm, as drawn, 99.9+%
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Magnesium, microfoil, 50x50mm, thinness 1.0μm, specific density 174μg/cm2, 6 micron aluminum permanent support, 99.9%
Magnesium, wire reel, 2m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, microleaf, 50x50mm, thinness 1.0μm, specific density 174μg/cm2, removable support, 99.9%
Magnesium, wire reel, 2m, diameter 1.0mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 0.125mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 0.4mm, as drawn, 99.9+%
Magnesium, wire reel, 1m, diameter 0.4mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 0.25mm, as drawn, 99.9+%