Merck
  • Characterization of a cDNA from Beta maritima that confers nickel tolerance in yeast.
Characterization of a cDNA from Beta maritima that confers nickel tolerance in yeast.
Gene (2014-02-04)
Gonensin O Bozdag, Alaattin Kaya, Ahmet Koc, Gundula A Noll, Dirk Prüfer, Hüseyin Caglar Karakaya
ABSTRACT
Nickel is an essential micronutrient due to its involvement in many enzymatic reactions as a cofactor. However, excess of this element is toxic to biological systems. Here, we constructed a cDNA library from Beta maritima and screened it in the yeast system to identify genes that confer resistance to toxic levels of nickel. A cDNA clone (NIC6), which encodes for a putative membrane protein with unknown function, was found to help yeast cells to tolerate toxic levels of nickel. A GFP fused form of Nic6 protein was localized to multivesicular structures in tobacco epidermal cells. Thus, our results suggest a possible role of Nic6 in nickel and intracellular ion homeostasis.
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Nickel, rod, 200mm, diameter 40mm, 99%
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Nickel, wire reel, 5m, diameter 0.5mm, as drawn, 99%
Nickel, wire reel, 2m, diameter 0.75mm, as drawn, 99.99+%
Nickel, wire reel, 500m, diameter 0.125mm, hard, 99.98%
Nickel, foil, 6mm disks, thickness 0.007mm, 99.95%
Nickel, wire reel, 5m, diameter 0.25mm, as drawn, 99.99+%
Nickel, wire reel, 5m, diameter 0.5mm, hard, 99.98%
Nickel, wire reel, 5m, diameter 0.05mm, as drawn, 99.99+%
Nickel, foil, 6mm disks, thickness 0.015mm, 99.9%
Nickel, foil, 6mm disks, thickness 0.05mm, annealed, 99%
Nickel, wire reel, 500m, diameter 0.25mm, hard, 99.98%
Nickel, foil, not light tested, 100x100mm, thickness 0.002mm, 99.95%
Nickel, foil, not light tested, 25x25mm, thickness 0.001mm, 99.95%
Nickel, foil, 150x300mm, thickness 0.25mm, annealed, 99.98%
Nickel, pellets, 50g, max. size 5mm, 99.99+%
Nickel, wire reel, 0.5m, diameter 0.05mm, as drawn, 99.99+%
Nickel, foil, 4mm disks, thickness 0.02mm, 99.9%
Nickel, foil, 0.5m coil, thickness 0.038mm, 99.9%
Nickel, foil, 0.2m coil, thickness 0.004mm, hard, 99%
Nickel, foil, 150x300mm, thickness 0.5mm, annealed, 99.98%
Nickel, foil, 10mm disks, thickness 0.0125mm, 99.9%
Nickel, foil, 0.5m coil, thickness 0.15mm, coil width 12mm, annealed, 99%
Nickel, foil, 50x50mm, thickness 1.0mm, annealed, 99%
Nickel, foil, 25x25mm, thickness 3.2mm, as rolled, 99.99+%