Merck
  • Plasmon-assisted photocurrent generation from silver nanoparticle monolayers combined with porphyrins via their different chain-length alkylcarboxylates.
Plasmon-assisted photocurrent generation from silver nanoparticle monolayers combined with porphyrins via their different chain-length alkylcarboxylates.
Journal of nanoscience and nanotechnology (2014-04-18)
Takayoshi Kakuta, Hiroki Kon, Azusa Kajikawa, Katsuhiko Kanaizuka, Shigeta Yagyu, Ryosuke Miyake, Manabu Ishizakil, Keirei Uruma, Takanari Togashi, Masatomi Sakamoto, Masato Kurihara
ABSTRACT
Three-typed porphyrin derivatives with a different chain-length alkylcarboxylic acid as their peripheral anchor group have been prepared. Anodic photocurrents were observed in a simple system where the porphyrin derivatives were directly anchored on an indium tin oxide (ITO) electrode. Cathodic photocurrents and their plasmon-assisted enhancement appeared from an Ag nanoparticle (Ag NP) composite monolayer combined with the porphyrin derivatives on the ITO electrode. In the photocurrent generation mechanism, Ag NPs played both the roles as photon- and energy-transfer to the porphyrin derivatives. The plasmon-assisted enhancement was affected by the chain-lengths of the peripheral anchor groups.
Sigma-Aldrich
Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥10 U/mg
Sigma-Aldrich
Silver nanowires, diam. × L 50 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver, conductive paste
Sigma-Aldrich
Esterase Isoenzyme 1 porcine liver, recombinant, recombinant, expressed in E. coli, ≥30.0 U/g
Sigma-Aldrich
Silver, colloidal, 65-75% Ag basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, powder, <250 μm, 99.99% trace metals basis
Sigma-Aldrich
Silver, wool, diam. 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Esterase from porcine liver, lyophilized, powder, slightly beige, ≥50 U/mg
Sigma-Aldrich
Esterase from rabbit liver, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Esterase from porcine liver, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Esterase from porcine liver, ammonium sulfate suspension, ≥150 units/mg protein (biuret)
Sigma-Aldrich
Silver nanowires, diam. × L 100 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Silver, foil, 10mm disks, thickness 0.5mm, as rolled, 99.99+%
Silver, wire reel, 1m, diameter 0.025mm, as drawn, 99.99%
Silver, wire reel, 20m, diameter 0.125mm, annealed, 99.99%
Silver, foil, 50x50mm, thickness 0.25mm, as rolled, 99.95+%
Silver, wire reel, 1m, diameter 0.050mm, gold plated, 99.99%
Silver, foil, 0.5m coil, thickness 0.15mm, as rolled, 99.95+%
Silver, foil, 10mm disks, thickness 0.008mm, 99.95+%
Silver, foil, 10mm disks, thickness 1.0mm, as rolled, 99.99+%
Silver, foil, 10mm disks, thickness 0.0125mm, 99.95+%
Silver, wire reel, 0.5m, diameter 0.25mm, as drawn, 99.997%
Silver, foil, 10mm disks, thickness 0.05mm, as rolled, 99.95+%