Merck
  • The use of transdermal buprenorphine to relieve radiotherapy-related pain in head and neck cancer patients.
The use of transdermal buprenorphine to relieve radiotherapy-related pain in head and neck cancer patients.
Cancer investigation (2013-06-14)
Johan Menten, Isabelle Carpentier, Harlinde Deschutter, Sandra Nuyts, Karen Van Beek
ABSTRACT
Many head and neck cancer (HNC) patients experience painful therapy-related mucositis and dermatitis. This prospective observational study evaluated transdermal buprenorphine use in HNC patients to relieve treatment-related pain. During treatment with paracetamol or tramadol, visual analogue scale (VAS)-pain scores >30/100 occurred in 26/45 patients 4 weeks after starting cancer therapy, persisting for ≥2 weeks after treatment. These patients subsequently received transdermal buprenorphine. Pain therapy should be more accurately up-titrated to the maximum recommended dose (140 μg/hr) where necessary to maintain pain scores ≤30/100 and, for some patients, should be continued for 6 weeks after the last cancer treatment day.
Product Description
Supelco
Buprenorphine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Buprenorphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Buprenorphine hydrochloride solution, analytical standard, for drug analysis
Sigma-Aldrich
Buprenorphine hydrochloride