Merck
  • Mg2+ regulates cytotoxic functions of NK and CD8 T cells in chronic EBV infection through NKG2D.
Mg2+ regulates cytotoxic functions of NK and CD8 T cells in chronic EBV infection through NKG2D.
Science (New York, N.Y.) (2013-07-13)
Benjamin Chaigne-Delalande, Feng-Yen Li, Geraldine M O'Connor, Marshall J Lukacs, Ping Jiang, Lixin Zheng, Amber Shatzer, Matthew Biancalana, Stefania Pittaluga, Helen F Matthews, Timothy J Jancel, Jack J Bleesing, Rebecca A Marsh, Taco W Kuijpers, Kim E Nichols, Carrie L Lucas, Sunil Nagpal, Huseyin Mehmet, Helen C Su, Jeffrey I Cohen, Gulbu Uzel, Michael J Lenardo
ABSTRACT
The magnesium transporter 1 (MAGT1) is a critical regulator of basal intracellular free magnesium (Mg(2+)) concentrations. Individuals with genetic deficiencies in MAGT1 have high levels of Epstein-Barr virus (EBV) and a predisposition to lymphoma. We show that decreased intracellular free Mg(2+) causes defective expression of the natural killer activating receptor NKG2D in natural killer (NK) and CD8(+) T cells and impairs cytolytic responses against EBV. Notably, magnesium supplementation in MAGT1-deficient patients restores intracellular free Mg(2+) and NKG2D while concurrently reducing EBV-infected cells in vivo, demonstrating a link between NKG2D cytolytic activity and EBV antiviral immunity in humans. Moreover, these findings reveal a specific molecular function of free basal intracellular Mg(2+) in eukaryotic cells.
Sigma-Aldrich
Magnesium preparation, Highly Reactive Rieke® Metal, suspension, 2.5 g in THF
Sigma-Aldrich
Magnesium, turnings, 5-25 mm, 99.95% trace metals basis
Sigma-Aldrich
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Sigma-Aldrich
Magnesium, turnings, reagent grade, 98%
Sigma-Aldrich
Magnesium, ribbon, ≥99.0% Mg basis
Sigma-Aldrich
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Sigma-Aldrich
Magnesium, powder, ≥99%
Sigma-Aldrich
Magnesium, 20-230 mesh, reagent grade, 98%
Sigma-Aldrich
Magnesium, ReagentPlus®, ribbon, ≥99% trace metals basis
Sigma-Aldrich
Magnesium, grit, ≥99.0% (KT)
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Sigma-Aldrich
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Sigma-Aldrich
Magnesium, rod, diam. 6 mm, ≥99.9% trace metals basis