Merck
  • Restoration of primary anterior teeth using intracanal polyethylene fibers and composite: an in vivo study.
Restoration of primary anterior teeth using intracanal polyethylene fibers and composite: an in vivo study.
The journal of adhesive dentistry (2013-03-28)
Mahtab Memarpour, Fereshteh Shafiei
ABSTRACT
To evaluate the clinical performance of polyethylene fiber for anterior primary teeth restorations in children after 2.5 years, according to modified Ryge criteria. Fifty-five maxillary incisors with extensive caries in 24 Iranian patients (mean age 4.2 years) were restored. All teeth were treated endodontically. First, 4-mm layer of paste at the entrance of canal was removed, and a thin base layer of polycarboxylate cement was placed at the bottom of the prepared canal. All tooth surfaces were etched, rinsed and dried, and the dentin adhesive Single Bond (3M ESPE, St Paul, MN, USA) was applied. The teeth received intracanal fiber-reinforced resin (Ribbond Triaxial, Seattle, WA, USA) fixed with a flowable composite and the crowns were reconstructed with microhybrid composite resin (Z250, 3M). All restorations were evaluated every 6 months for 30 months by 2 independent evaluators. Statistical analysis was done with McNemar's test. The surface textures for most of the restorations were judged as excellent. There was no evidence of significant changes in marginal integrity. Most restored incisors (81%) received an Alpha rating for retention. The baseline and recall retention scores differed significantly (p=0.002). Polyethylene fiber posts associated with extensive composite restoration showed excellent clinical performance after 2.5 years in primary anterior teeth after pulp therapy.
Product Description
Polyethylene (LDPE), ERM®, certified reference material
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Supelco
Polyethylene, analytical standard, for GPC, 2,000
Sigma-Aldrich
Polyethylene, Medium density
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size