Skip to Content
Merck
All Photos(1)

Key Documents

ERMEC590

Polyethylene (LDPE)

ERM®, certified reference material

Synonym(s):

Polyethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(CH2CH2)nH
CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

application(s)

chemicals and industrial polymers

format

neat

storage temp.

−20°C

SMILES string

C=C

InChI

1S/C2H4/c1-2/h1-2H2

InChI key

VGGSQFUCUMXWEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
ERMEC590

Legal Information

ERM is a registered trademark of European Commission

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Thomas P J Linsinger et al.
Analytical chemistry, 81(10), 3792-3800 (2009-03-28)
The first reference materials certified for several polybrominated flame retardants in polymers were developed. Commercially available polyethylene and polypropylene were fortified with technical mixtures of Pentabrominated diphenylether (Penta-BDE), Octa-BDE, Deca-BDE, and Decabrominated biphenyl (BB) (where the capitalized forms refer to
Nina Bloecher et al.
Biofouling, 29(3), 237-246 (2013-02-27)
The hydroid Ectopleura larynx is one of the main fouling organisms on salmon aquaculture cages in Norway; this study investigated novel surface materials and microtopographies to deter its settlement. The settlement preferences of hydroid larvae for 12 materials with wettabilities
Thibault Honegger et al.
Lab on a chip, 13(8), 1538-1545 (2013-02-23)
In this paper, we introduce a dielectrophoresis (DEP)-based handling method that allows fine 3D manipulation of beads in suspension using a lab on a chip device. The device consists of two layers of linear electrodes on the top and bottom
Ryan M Baxter et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 101(3), 467-475 (2013-02-26)
Despite the widespread implementation of highly cross-linked polyethylene (HXLPE) liners to reduce the clinical incidence of osteolysis, it is not known if the improved wear resistance will outweigh the inflammatory potential of HXLPE wear debris generated in vivo. Thus, we
Maria C S Inacio et al.
Acta orthopaedica, 84(2), 145-152 (2013-03-15)
There is no substantial clinical evidence for the superiority of alternative bearings in total knee arthroplasty (TKA). We compared the short-term revision risk in alternative surface bearing knees (oxidized zirconium (OZ) femoral implants or highly crosslinked polyethylene (HXLPE) inserts) with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service