Merck
  • Characterization of the diatomite binding domain in the ribosomal protein L2 from E. coli and functions as an affinity tag.
Characterization of the diatomite binding domain in the ribosomal protein L2 from E. coli and functions as an affinity tag.
Applied microbiology and biotechnology (2012-08-29)
Junhua Li, Yang Zhang, Yanjun Yang
ABSTRACT
The ribosomal protein L2, a constituent protein of the 50S large ribosomal subunit, can be used as Si-tag using silica particles for the immobilization and purification of recombinant proteins (Ikeda et al. (Protein Expr Purif 71:91-95, 2010); Taniguchi et al. (Biotechnol Bioeng 96:1023-1029, 2007)). We applied a diatomite powder, a sedimentary rock mainly composed with diatoms silica, as an affinity solid phase and small ubiquitin-like modifier (SUMO) technology to release a target protein from the solid phase. The L2 (203-273) was the sufficient region for the adsorption of ribosomal protein L2 on diatomite. We comparatively analyzed the different adsorption properties of the two deleted proteins of L2 (L2 (1-60, 203-273) and L2 (203-273)) on diatomite. The time required to reach adsorption equilibrium of L2 (203-273) fusion protein on diatomite was shorter than that of L2 (1-60, 203-273) fusion protein. The maximum adsorption capacity of L2 (203-273) fusion protein was larger than that of L2 (1-60, 203-273) fusion protein. In order to study whether the L2 (203-273) can function as an affinity purification tag, SUMO was introduced as one specific protease cleavage site between the target protein and the purification tags. The L2 (203-273) and SUMO fusion protein purification method was tested using enhanced green fluorescent protein as a model protein; the result shows that the purification performance of this affinity purification method was good. The strong adsorption characteristic of L2 (203-273) on diatomite also provides a potential protein fusion tag for the immobilization of enzyme.
Product Description
Supelco
Celite® 545 AW, reagent grade
Supelco
Celatom®, acid-washed, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
Celite® 503, filter aid, treated with sodium carbonate, calcined
Sigma-Aldrich
Celpure® P65, meets USP/NF testing specifications
Sigma-Aldrich
Hyflo® Super Cel®, filter aid, flux calcined, treated with sodium carbonate
Sigma-Aldrich
Celite® S, filter aid, dried, untreated
Sigma-Aldrich
Diatomaceous earth non-washed, powder
Sigma-Aldrich
Diatomaceous earth
Sigma-Aldrich
Filter agent, Celite® 545
Sigma-Aldrich
Celpure® P1000, meets USP/NF testing specifications
Sigma-Aldrich
Celite® 545, filter aid, treated with sodium carbonate, flux calcined
Sigma-Aldrich
Celite® 545 AW, filter aid, acid washed, treated with sodium carbonate, flux calcined