Skip to Content
Merck
All Photos(2)

Documents

392545

Sigma-Aldrich

Diatomaceous earth

Synonym(s):

Diatomaceous earth, flux-calcined

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
11101527
PubChem Substance ID:
NACRES:
NA.21

form

powder

particle size

150 mesh (<=12%, Tyler sieve retain)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diatomaceous earth is natural amorphous silica formed from the fossilized skeletons of diatoms. It has high water and oil absorption ability. The potential toxicity of different samples of DE has been assessed.

Application

Diatomaceous earth may be used as an adsorbent for column chromatography.
It may be used in the synthesis of the following:
  • Chitosan modified DE for the abstraction of humic acid from raw water.
  • High purity solar grade silicon (SOG-Si).
  • Porous silica ceramics.
  • Diatomite-based composite with enhanced sorption characteristics.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of novel diatomite-based composites: applications in organic ef?uents sorption.
Khelifa N, et al.
Desalination and Water Treatment, 1-10 (2015)
Porous ceramic monoliths based on diatomite.
Saponjic A, et al.
Ceramics International, 41(8), 9745-9752 (2015)
High purity silicon materials prepared through wet-chemical and electrochemical approaches.
Homma T, et al.
Electrochimica Acta, 179, 512-518 (2015)
Study on Adsorption Removal of Humic Acid in Water by Modified Clays.
Wang YH, et al.
Applied Mechanics and Materials, 707, 247-250 (2015)
The global variability of diatomaceous earth toxicity: a physicochemical and in vitro investigation.
Nattrass C, et al.
Journal of Occupational Medicine and Toxicology (London, England), 10(1), 23-23 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service