Merck
Dreaming without REM sleep.
Consciousness and cognition (2012-06-01)
Delphine Oudiette, Marie-José Dealberto, Ginevra Uguccioni, Jean-Louis Golmard, Milagros Merino-Andreu, Mehdi Tafti, Lucile Garma, Sophie Schwartz, Isabelle Arnulf
ABSTRACT
To test whether mental activities collected from non-REM sleep are influenced by REM sleep, we suppressed REM sleep using clomipramine 50mg (an antidepressant) or placebo in the evening, in a double blind cross-over design, in 11 healthy young men. Subjects were awakened every hour and asked about their mental activity. The marked (81%, range 39-98%) REM-sleep suppression induced by clomipramine did not substantially affect any aspects of dream recall (report length, complexity, bizarreness, pleasantness and self-perception of dream or thought-like mentation). Since long, complex and bizarre dreams persist even after suppressing REM sleep either partially or totally, it suggests that the generation of mental activity during sleep is independent of sleep stage.
Sigma-Aldrich
Clomipramine hydrochloride, ≥98% (HPLC), powder
Supelco
Clomipramine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®