Merck
  • Comparison of n-tetradecane/electrolyte emulsions properties stabilized by DPPC and DPPC vesicles in the electrolyte solution.
Comparison of n-tetradecane/electrolyte emulsions properties stabilized by DPPC and DPPC vesicles in the electrolyte solution.
Colloids and surfaces. B, Biointerfaces (2010-12-07)
Agnieszka Ewa Wiącek
ABSTRACT
The properties of n-tetradecane/electrolyte emulsions with DPPC or DPPC vesicles in the electrolyte solution were investigated. The DPPC molecules form different aggregates, which possess different surface affinity, size and structure, and therefore we assumed some differences in the adsorption at the oil droplet/water interface. The n-tetradecane emulsions in 1:1, 1:2 and 1:3 electrolytes were prepared by mechanical stirring in the presence of DPPC at natural pH. Electrokinetic properties of the systems were investigated taking into account the effective diameter and multimodal size distribution of the droplets as well as the zeta potentials using the dynamic light scattering technique. The zeta potential of the droplets was negative in all systems with NaCl. In the emulsions with CaCl(2) at a higher concentration of electrolyte and emulsions with LaCl(3) with all investigated concentrations, positive values were observed. Similar measurements were performed for DPPC vesicles in the electrolyte solution. The pH and ionic strength changes induce those in the electrical charge of DPPC layer or vesicle surface. This is due to the fact that the DPPC molecule contains -PO(-) and -N(CH(3))(3) groups, which are in equilibrium with H(+) and OH(-), as well as other ions present in the solution, i.e. Na(+), Ca(2+), La(3+) or Cl(-). In the n-tetradecane/electrolyte emulsion stabilized by DPPC or DPPC vesicles the zeta potential may be also related to acid-base interactions. The effect of the ions from the solution on the DPPC layer adsorbed on n-tetradecane droplets or DPPC vesicles is discussed.
Sigma-Aldrich
Tetradecane, ≥99%
Sigma-Aldrich
Tetradecane, olefin free, ≥99.0% (GC)
Supelco
Tetradecane, analytical standard