Merck
  • Large-scale 13C-flux analysis reveals distinct transcriptional control of respiratory and fermentative metabolism in Escherichia coli.
Large-scale 13C-flux analysis reveals distinct transcriptional control of respiratory and fermentative metabolism in Escherichia coli.
Molecular systems biology (2011-04-01)
Bart R B Haverkorn van Rijsewijk, Annik Nanchen, Sophie Nallet, Roelco J Kleijn, Uwe Sauer
ABSTRACT
Despite our increasing topological knowledge on regulation networks in model bacteria, it is largely unknown which of the many co-occurring regulatory events actually control metabolic function and the distribution of intracellular fluxes. Here, we unravel condition-dependent transcriptional control of Escherichia coli metabolism by large-scale (13)C-flux analysis in 91 transcriptional regulator mutants on glucose and galactose. In contrast to the canonical respiro-fermentative glucose metabolism, fully respiratory galactose metabolism depends exclusively on the phosphoenol-pyruvate (PEP)-glyoxylate cycle. While 2/3 of the regulators directly or indirectly affected absolute flux rates, the partitioning between different pathways remained largely stable with transcriptional control focusing primarily on the acetyl-CoA branch point. Flux distribution control was achieved by nine transcription factors on glucose, including ArcA, Fur, PdhR, IHF A and IHF B, but was exclusively mediated by the cAMP-dependent Crp regulation of the PEP-glyoxylate cycle flux on galactose. Five further transcription factors affected this flux only indirectly through cAMP and Crp by increasing the galactose uptake rate. Thus, E. coli actively limits its galactose catabolism at the expense of otherwise possible faster growth.
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide with 1% tert-Butyldimethylchlorosilane, ≥95%
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide-d, 98 atom % D