Merck
  • Mushroom toxicosis in dogs in general practice causing gastroenteritis, ptyalism and elevated serum lipase activity.
Mushroom toxicosis in dogs in general practice causing gastroenteritis, ptyalism and elevated serum lipase activity.
The Journal of small animal practice (2013-02-13)
J Hall, L Barton
ABSTRACT
Mushroom toxicosis is rarely diagnosed in dogs and is poorly reported in the veterinary literature. This report suggests that mushroom toxicosis is a potentially under-diagnosed condition in first opinion practice in the UK. Nine dogs with clinical signs consistent with mushroom toxicosis were identified from the records of an out-of-hours emergency service between August 2010 and January 2011. Four dogs were later excluded because of clinical inconsistencies. Clinical signs included acute profuse ptyalism (5/5), diarrhoea (5/5), vomiting (4/5), hypovolaemia (4/5), stuporous (3/5) or obtunded mentation (1/5), miosis (2/5) and hypothermia (2/5). Serum lipase activity was elevated in 4/4 dogs; canine-specific pancreatic lipase was elevated in the remaining dog. Four dogs recovered with aggressive intravenous fluid therapy, analgesia and supportive care; the remaining dog was euthanased due to severe clinical signs and financial constraints. Mushroom toxicosis is an important differential diagnosis for acute gastroenteritis and one possible cause of some cases of "Seasonal Canine Illness". Affected dogs may demonstrate elevated pancreatic enzymes and mushroom toxicosis should be considered in cases of elevated lipase or abnormal semi-quantitative canine-specific pancreatic lipase activities.
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Aspergillus oryzae, solution, ≥100,000 U/g, white, beige
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg