Skip to Content
Merck
All Photos(1)

Key Documents

L9031

Sigma-Aldrich

Lipase from Mucor miehei

lyophilized powder, ≥4,000 units/mg solid (using olive oil)

Synonym(s):

Triacylglycerol acylhydrolase, Triacylglycerol lipase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

form

lyophilized powder

Quality Level

specific activity

≥4,000 units/mg solid (using olive oil)

storage temp.

2-8°C

InChI

1S/C11H9N3O2.Na/c15-8-4-5-9(10(16)7-8)13-14-11-3-1-2-6-12-11;/h1-7,16H,(H,12,14);/q;+1/b13-9-;

InChI key

QWZUIMCIEOCSJF-CHHCPSLASA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Lipase is produced by several animals, microbes, and plants. Lipase from Mucor sp. is a fungal lipase.[1] It is a member of the serine hydrolase family.[2]

Application

Lipase from Mucor miehei can be used to drive the synthesis of flavor esters. [3]
Lipase from Mucor miehei has been used in a study to assess the nonthermal effect of microwave irradiation in nonaqueous enzymatic esterification. [4] It has also been used in a study to investigate the hydrolysis of virgin coconut oil using immobilized lipase in a batch reactor. [5]
Lipases are used industrially for the resolution of chiral compounds and the transesterification production of biodiesel.

Biochem/physiol Actions

Mucor miehei lipase hydrolizes triglycerides and catalyzes the synthesis of esters from glycerol and long-chain fatty acids in vitro. Tri-, di-, and monoglycerides are hydrolyzed (in decreasing order of rate).
Microbial lipase has a wide industrial application due to its stability, broad substrate specificity, and selectivity.[1] It may be used in the synthesis of biosurfactants and finds application in organic chemical processing, dairy industry, oleochemical industry, detergent formulations, paper, cosmetics, nutrition, and the pharmaceutical industry.[6]
Tri-, di-, and monoglycerides are hydrolyzed (in decreasing order of rate).

Unit Definition

One unit will hydrolyze 1.0 microequivalent of fatty acid from a triglyceride in 1 hr at pH 7.7 at 37 °C using olive oil.

Analysis Note

Protein determined by biuret.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lee Suan Chua et al.
Enzyme research, 2012, 542589-542589 (2012-09-07)
Hydrolysis of virgin coconut oil (VCO) had been carried out by using an immobilised lipase from Mucor miehei (Lipozyme) in a water-jacketed batch reactor. The kinetic of the hydrolysis was investigated by varying the parameters such as VCO concentration, enzyme
Hui-da Wan et al.
Applied biochemistry and biotechnology, 166(6), 1454-1462 (2012-01-21)
Microwave has nonthermal effects on enzymatic reactions, mainly caused by the polarities of the solvents and substrates. In this experiment, a model reaction with caprylic acid and butanol that was catalyzed by lipase from Mucor miehei in alkanes or arenes
Agnieszka Stobiecka
Journal of photochemistry and photobiology. B, Biology, 80(1), 9-18 (2005-06-21)
Steady-state and time-resolved fluorescence-quenching measurements have been performed to study multitryptophan lipase from filamentous fungus Rhizomucor miehei. Using the steady-state acrylamide fluorescence quenching data and the fluorescence-quenching-resolved-spectra (FQRS) method, the total emission spectrum of native ("closed-lid") lipase has been decomposed
H S Ryu et al.
Applied microbiology and biotechnology, 70(3), 321-326 (2005-08-10)
A Photobacterium strain, M37, showing lipolytic activity, was previously isolated from an intertidal flat of the Yellow Sea in Korea and identified as Photobacterium lipolyticum sp. nov. In the present study, the corresponding gene was cloned using the shotgun method.
R Sharma et al.
Biotechnology advances, 19(8), 627-662 (2003-10-11)
Lipases (triacylglycerol acylhydrolases, EC 3.1.1.3) catalyze the hydrolysis and the synthesis of esters formed from glycerol and long-chain fatty acids. Lipases occur widely in nature, but only microbial lipases are commercially significant. The many applications of lipases include speciality organic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service