Merck
  • Characterization of in vitro protein oxidation using mass spectrometry: a time course study of oxidized alpha-amylase.
Characterization of in vitro protein oxidation using mass spectrometry: a time course study of oxidized alpha-amylase.
Archives of biochemistry and biophysics (2012-12-19)
André M N Silva, Susana L Marçal, Rui Vitorino, Maria R M Domingues, Pedro Domingues
ABSTRACT
The study of protein damage by oxidative processes and its influence on protein activity is central to understanding the deleterious effects of oxidative stress on biological systems. This paper will focus on the study of enzyme inactivation by oxidative modifications, utilizing α-amylase from Bacillus species. (BAA) as a model protein. Oxidative stress was induced using metal catalyzed oxidation (MCO). The enzymatic activity of BAA was correlated with the oxidative damage induced to the protein. Off-line nano-HPLC-MALDI-TOF/TOF-MS was used to characterize the oxidative modifications occurring to the protein. Additionally, semi-quantitative analysis was employed in order to evaluate the significance of the various oxidative modifications. BAA oxidation was found to be deleterious to its enzymatic activity. A total of 10 amino acid residues were found to have an oxidation degree above 50%, out of which eight were methionine and tryptophan. Residues in the proximity of key structural elements were found to be particularly susceptible to oxidation. The oxidative process was found to be governed by the nature of the amino acid residues side chain and, to a lesser extent, their location within the three dimensional structure of the protein.
Sigma-Aldrich
α-Amylase from porcine pancreas, PMSF Treated, Type I-A, saline suspension, ≥1000 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, ≥400 units/mg protein (Lowry)
Sigma-Aldrich
α-Amylase from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase, heat-stable, solution, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
α-Amylase from porcine pancreas, Type I-A, PMSF treated, saline suspension, 700-1400 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~380 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~50 U/mg
Sigma-Aldrich
Taka-Diastase from Aspergillus oryzae, powder, slightly beige, ~100 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., liquid
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, ≥150 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from Bacillus amyloliquefaciens, liquid, ≥250 units/g
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, aqueous solution, ≥800 FAU/g
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~30 U/mg
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)
Supelco
α-Amylase from Bacillus licheniformis, suitable for determination of starch (Kit STA-20)
Sigma-Aldrich
α-Amylase from human saliva, Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, Type XII-A, saline solution, ≥500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from human saliva, Type XIII-A, lyophilized powder, 300-1,500 units/mg protein
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~1.5 U/mg (~0.2 U acc. to Willstätter)