Merck
  • LncRNA THUMPD3-AS1 promotes invasion and EMT in gastric cancer by regulating the miR-1297/BCAT1 pathway.
LncRNA THUMPD3-AS1 promotes invasion and EMT in gastric cancer by regulating the miR-1297/BCAT1 pathway.
iScience (2023-09-14)
Zaibo Zhang, Yong Li, Liqiao Fan, Bingyu Wang, Wenbo Liu, Jiaxiang Cui, Bibo Tan
ABSTRACT
Long noncoding RNA (lncRNA) plays crucial roles in the development of gastric cancer (GC); however, studies of their mechanisms of action are needed to determine their clinical value. The aim of this study is to explore the effects and mechanisms of THUMPD3-AS1 in GC. Elevated levels of THUMPD3-AS1 were observed in GC and demonstrated a significant positive correlation with poor prognosis. Functionally, THUMPD3-AS1 promoted GC cell proliferation, migration, invasion, and epithelial-mesenchymal transition (EMT) and induced tumor growth in vivo. THUMPD3-AS1 exerts its regulatory function on BCAT1 through competitive binding with miR-1297. Further investigations confirmed that both THUMPD3-AS1 and miR-1297 interact with BCAT1. These findings suggest that THUMPD3-AS1 promotes GC invasion and EMT by regulating the miR-1297/BCAT1 pathway, indicating that THUMPD3-AS1 may serve as a biomarker and therapeutic target for GC.
Sigma-Aldrich
Monoclonal Anti-Calbindin-D-28K antibody produced in mouse, clone CB-955, ascites fluid
Sigma-Aldrich
Monoclonal Anti-BCAT1 antibody produced in mouse, clone 1F8, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-CDH1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse